Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63239197-63239466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC7
Entrez Gene ID
79444
Gene Name
baculoviral IAP repeat containing 7
Gene Aliases
KIAP, LIVIN, ML-IAP, MLIAP, RNF50
UniGene
Hs.256126
Gene Chromosome Location
Chr.20: 63235883-63240507 on Build GRCh38
Transcripts
NM_022161.3 NM_139317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7152020 c.745G>A p.E249K Chr.20: 63239453-63239453 on Build GRCh38
5513645 c.753G>A p.T251T Chr.20: 63239461-63239461 on Build GRCh38
7084744 c.681G>A p.A227A Chr.20: 63239389-63239389 on Build GRCh38
7123731 c.699C>A p.P233P Chr.20: 63239407-63239407 on Build GRCh38
444199 c.667G>C p.E223Q Chr.20: 63239375-63239375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765257076 Chr.20: 63239325-63239325 on Build GRCh38
rs778215156 Chr.20: 63239247-63239248 on Build GRCh38
rs758964512 Chr.20: 63239219-63239219 on Build GRCh38
rs757458592 Chr.20: 63239399-63239399 on Build GRCh38
rs560298175 Chr.20: 63239224-63239224 on Build GRCh38
rs150789557 Chr.20: 63239431-63239431 on Build GRCh38
rs535809908 Chr.20: 63239371-63239371 on Build GRCh38
rs775098582 Chr.20: 63239291-63239291 on Build GRCh38
rs752719404 Chr.20: 63239339-63239339 on Build GRCh38
rs1040161389 Chr.20: 63239293-63239293 on Build GRCh38
rs776545168 Chr.20: 63239310-63239310 on Build GRCh38
rs771535832 Chr.20: 63239283-63239283 on Build GRCh38
rs200528640 Chr.20: 63239329-63239329 on Build GRCh38
rs756386842 Chr.20: 63239340-63239340 on Build GRCh38
rs756265998 Chr.20: 63239278-63239278 on Build GRCh38
rs775260228 Chr.20: 63239229-63239229 on Build GRCh38
rs61737394 Chr.20: 63239439-63239439 on Build GRCh38
rs373757098 Chr.20: 63239389-63239389 on Build GRCh38
rs747763479 Chr.20: 63239441-63239441 on Build GRCh38
rs768540333 Chr.20: 63239300-63239302 on Build GRCh38
rs557505715 Chr.20: 63239374-63239374 on Build GRCh38
rs901495544 Chr.20: 63239294-63239294 on Build GRCh38
rs776898586 Chr.20: 63239267-63239282 on Build GRCh38
rs758907331 Chr.20: 63239362-63239362 on Build GRCh38
rs766291917 Chr.20: 63239386-63239386 on Build GRCh38
rs981460611 Chr.20: 63239377-63239377 on Build GRCh38
rs760352384 Chr.20: 63239238-63239238 on Build GRCh38
rs193196289 Chr.20: 63239330-63239330 on Build GRCh38
rs564528998 Chr.20: 63239304-63239304 on Build GRCh38
rs374132984 Chr.20: 63239430-63239430 on Build GRCh38
rs869203806 Chr.20: 63239254-63239255 on Build GRCh38
rs199537462 Chr.20: 63239387-63239387 on Build GRCh38
rs141418586 Chr.20: 63239418-63239418 on Build GRCh38
rs376332500 Chr.20: 63239438-63239438 on Build GRCh38
rs376620293 Chr.20: 63239353-63239353 on Build GRCh38
rs558241909 Chr.20: 63239396-63239396 on Build GRCh38
rs895643525 Chr.20: 63239407-63239407 on Build GRCh38
rs767591214 Chr.20: 63239446-63239448 on Build GRCh38
rs761821260 Chr.20: 63239251-63239251 on Build GRCh38
rs750757399 Chr.20: 63239420-63239420 on Build GRCh38
rs367617870 Chr.20: 63239285-63239285 on Build GRCh38
rs3215649 Chr.20: 63239253-63239254 on Build GRCh38
rs780842743 Chr.20: 63239425-63239425 on Build GRCh38
rs757380953 Chr.20: 63239244-63239244 on Build GRCh38
rs1075556 Chr.20: 63239282-63239282 on Build GRCh38
rs768132359 Chr.20: 63239307-63239307 on Build GRCh38
rs771892125 Chr.20: 63239228-63239228 on Build GRCh38
rs773991334 Chr.20: 63239341-63239341 on Build GRCh38
rs761504955 Chr.20: 63239311-63239311 on Build GRCh38
rs779286324 Chr.20: 63239354-63239354 on Build GRCh38
rs1040130599 Chr.20: 63239448-63239448 on Build GRCh38
rs746405681 Chr.20: 63239361-63239361 on Build GRCh38
rs914849161 Chr.20: 63239234-63239234 on Build GRCh38
rs985604368 Chr.20: 63239319-63239319 on Build GRCh38
rs765068567 Chr.20: 63239253-63239253 on Build GRCh38
rs954369067 Chr.20: 63239317-63239317 on Build GRCh38
rs1077019 Chr.20: 63239375-63239375 on Build GRCh38
rs201147952 Chr.20: 63239267-63239267 on Build GRCh38
rs151009486 Chr.20: 63239334-63239334 on Build GRCh38
rs201155031 Chr.20: 63239442-63239442 on Build GRCh38
rs752950376 Chr.20: 63239276-63239276 on Build GRCh38
rs759028214 Chr.20: 63239424-63239424 on Build GRCh38
rs4637179 Chr.20: 63239260-63239260 on Build GRCh38
rs773213702 Chr.20: 63239448-63239450 on Build GRCh38
rs781203230 Chr.20: 63239271-63239271 on Build GRCh38
rs148286398 Chr.20: 63239388-63239388 on Build GRCh38
rs397840121 Chr.20: 63239256-63239257 on Build GRCh38
rs762895747 Chr.20: 63239378-63239378 on Build GRCh38
rs761721684 Chr.20: 63239360-63239360 on Build GRCh38
rs764218395 Chr.20: 63239350-63239350 on Build GRCh38
rs771730459 Chr.20: 63239290-63239290 on Build GRCh38
rs764010820 Chr.20: 63239243-63239243 on Build GRCh38
rs768419850 Chr.20: 63239372-63239372 on Build GRCh38
rs869270953 Chr.20: 63239257-63239258 on Build GRCh38
rs745419633 Chr.20: 63239287-63239287 on Build GRCh38
rs746077865 Chr.20: 63239293-63239296 on Build GRCh38

View Full Product Details