Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39661340-39661569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT5
Entrez Gene ID
10317
Gene Name
beta-1,3-galactosyltransferase 5
Gene Aliases
B3GalT-V, B3GalTx, B3T5, GLCT5, beta-1,3-GalTase 5, beta-3-Gx-T5, beta3Gal-T5
UniGene
Hs.733638
Gene Chromosome Location
Chr.21: 39612940-39662889 on Build GRCh38
Transcripts
NM_001278650.1 NM_006057.2 NM_033170.2 NM_033171.2 NM_033172.2 XM_017028227.1 XM_017028228.1 XM_017028229.1 XM_017028230.1 XM_017028231.1 XM_017028232.1 XM_017028233.1 XM_017028234.1 XM_017028235.1 XM_017028236.1 XM_017028237.1 XM_017028238.1 XM_017028239.1 XM_017028240.1 XM_017028241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7338269 c.794C>T p.T265I Chr.21: 39661353-39661353 on Build GRCh38
4562576 c.932G>A p.*311* Chr.21: 39661491-39661491 on Build GRCh38
3912120 c.843C>T p.I281I Chr.21: 39661402-39661402 on Build GRCh38
4664862 c.907C>T p.R303W Chr.21: 39661466-39661466 on Build GRCh38
5718136 c.802C>T p.P268S Chr.21: 39661361-39661361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200876589 Chr.21: 39661418-39661418 on Build GRCh38
rs1019871844 Chr.21: 39661390-39661390 on Build GRCh38
rs373479020 Chr.21: 39661467-39661467 on Build GRCh38
rs758280614 Chr.21: 39661515-39661515 on Build GRCh38
rs74843485 Chr.21: 39661544-39661544 on Build GRCh38
rs139601001 Chr.21: 39661436-39661436 on Build GRCh38
rs372024763 Chr.21: 39661425-39661425 on Build GRCh38
rs762547834 Chr.21: 39661463-39661464 on Build GRCh38
rs747666429 Chr.21: 39661542-39661542 on Build GRCh38
rs368740513 Chr.21: 39661463-39661463 on Build GRCh38
rs757514283 Chr.21: 39661472-39661472 on Build GRCh38
rs1026987085 Chr.21: 39661457-39661457 on Build GRCh38
rs766898271 Chr.21: 39661382-39661382 on Build GRCh38
rs376663833 Chr.21: 39661428-39661428 on Build GRCh38
rs968994929 Chr.21: 39661545-39661545 on Build GRCh38
rs752802027 Chr.21: 39661403-39661403 on Build GRCh38
rs764255566 Chr.21: 39661482-39661482 on Build GRCh38
rs1045392011 Chr.21: 39661435-39661435 on Build GRCh38
rs771791255 Chr.21: 39661363-39661363 on Build GRCh38
rs151007676 Chr.21: 39661406-39661406 on Build GRCh38
rs746544737 Chr.21: 39661372-39661372 on Build GRCh38
rs745684366 Chr.21: 39661426-39661426 on Build GRCh38
rs867063591 Chr.21: 39661380-39661380 on Build GRCh38
rs771597548 Chr.21: 39661465-39661465 on Build GRCh38
rs200631873 Chr.21: 39661398-39661398 on Build GRCh38
rs373768451 Chr.21: 39661514-39661514 on Build GRCh38
rs556149221 Chr.21: 39661474-39661474 on Build GRCh38
rs372655965 Chr.21: 39661381-39661381 on Build GRCh38
rs768270855 Chr.21: 39661466-39661467 on Build GRCh38
rs774654350 Chr.21: 39661466-39661466 on Build GRCh38
rs768323326 Chr.21: 39661454-39661454 on Build GRCh38
rs142843576 Chr.21: 39661370-39661370 on Build GRCh38
rs377437585 Chr.21: 39661505-39661505 on Build GRCh38
rs768270176 Chr.21: 39661373-39661373 on Build GRCh38
rs757212183 Chr.21: 39661424-39661424 on Build GRCh38
rs138903138 Chr.21: 39661464-39661464 on Build GRCh38
rs760852174 Chr.21: 39661477-39661477 on Build GRCh38
rs746639840 Chr.21: 39661453-39661453 on Build GRCh38
rs369617028 Chr.21: 39661513-39661513 on Build GRCh38
rs773923288 Chr.21: 39661374-39661374 on Build GRCh38
rs376207581 Chr.21: 39661534-39661534 on Build GRCh38
rs779684781 Chr.21: 39661519-39661519 on Build GRCh38
rs753952483 Chr.21: 39661483-39661483 on Build GRCh38
rs761818773 Chr.21: 39661485-39661485 on Build GRCh38
rs745606058 Chr.21: 39661360-39661360 on Build GRCh38
rs763524371 Chr.21: 39661376-39661376 on Build GRCh38
rs558391794 Chr.21: 39661538-39661538 on Build GRCh38
rs767783605 Chr.21: 39661402-39661402 on Build GRCh38
rs774801169 Chr.21: 39661393-39661393 on Build GRCh38
rs751307456 Chr.21: 39661533-39661533 on Build GRCh38

View Full Product Details