Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82058202-82058443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
UniGene
Hs.514599
Gene Chromosome Location
Chr.17: 82057872-82065821 on Build GRCh38
Transcripts
NM_022156.4 XM_005256393.2 XM_005256394.2 XM_006722288.2 XM_006722289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650254 c.1252C>T p.R418* Chr.17: 82058371-82058371 on Build GRCh38
1250791 c.1285C>T p.H429Y Chr.17: 82058252-82058252 on Build GRCh38
6398531 c.1235G>T p.R412L Chr.17: 82058388-82058388 on Build GRCh38
4070813 c.1253G>A p.R418Q Chr.17: 82058370-82058370 on Build GRCh38
7105478 c.1252C>G p.R418G Chr.17: 82058371-82058371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777832090 Chr.17: 82058362-82058362 on Build GRCh38
rs368394562 Chr.17: 82058297-82058297 on Build GRCh38
rs539331883 Chr.17: 82058386-82058386 on Build GRCh38
rs769221897 Chr.17: 82058395-82058395 on Build GRCh38
rs756770296 Chr.17: 82058270-82058270 on Build GRCh38
rs752467409 Chr.17: 82058420-82058420 on Build GRCh38
rs202067934 Chr.17: 82058417-82058417 on Build GRCh38
rs372300965 Chr.17: 82058313-82058313 on Build GRCh38
rs201246098 Chr.17: 82058316-82058316 on Build GRCh38
rs754277908 Chr.17: 82058278-82058279 on Build GRCh38
rs894661705 Chr.17: 82058399-82058399 on Build GRCh38
rs776826110 Chr.17: 82058305-82058305 on Build GRCh38
rs771072775 Chr.17: 82058304-82058304 on Build GRCh38
rs767025373 Chr.17: 82058273-82058273 on Build GRCh38
rs200669716 Chr.17: 82058321-82058321 on Build GRCh38
rs1056890591 Chr.17: 82058393-82058393 on Build GRCh38
rs1013204734 Chr.17: 82058263-82058263 on Build GRCh38
rs753620879 Chr.17: 82058287-82058287 on Build GRCh38
rs370082561 Chr.17: 82058317-82058317 on Build GRCh38
rs373844387 Chr.17: 82058278-82058278 on Build GRCh38
rs774994519 Chr.17: 82058396-82058396 on Build GRCh38
rs760845790 Chr.17: 82058246-82058246 on Build GRCh38
rs781445339 Chr.17: 82058372-82058372 on Build GRCh38
rs372912714 Chr.17: 82058410-82058410 on Build GRCh38
rs757631906 Chr.17: 82058302-82058303 on Build GRCh38
rs987514673 Chr.17: 82058308-82058308 on Build GRCh38
rs752597705 Chr.17: 82058345-82058345 on Build GRCh38
rs754769541 Chr.17: 82058334-82058334 on Build GRCh38
rs376659580 Chr.17: 82058370-82058370 on Build GRCh38
rs200336489 Chr.17: 82058265-82058265 on Build GRCh38
rs753464381 Chr.17: 82058414-82058414 on Build GRCh38
rs528501942 Chr.17: 82058332-82058332 on Build GRCh38
rs144652762 Chr.17: 82058249-82058249 on Build GRCh38
rs766750212 Chr.17: 82058248-82058248 on Build GRCh38
rs770981128 Chr.17: 82058371-82058371 on Build GRCh38
rs368733115 Chr.17: 82058389-82058389 on Build GRCh38
rs780010842 Chr.17: 82058282-82058282 on Build GRCh38
rs374612561 Chr.17: 82058292-82058292 on Build GRCh38
rs540579368 Chr.17: 82058223-82058223 on Build GRCh38
rs372697085 Chr.17: 82058322-82058322 on Build GRCh38
rs199929589 Chr.17: 82058228-82058228 on Build GRCh38
rs760319739 Chr.17: 82058397-82058397 on Build GRCh38
rs541870122 Chr.17: 82058333-82058333 on Build GRCh38
rs145282065 Chr.17: 82058374-82058374 on Build GRCh38
rs531072642 Chr.17: 82058302-82058302 on Build GRCh38
rs754799152 Chr.17: 82058288-82058288 on Build GRCh38
rs899380876 Chr.17: 82058424-82058424 on Build GRCh38
rs758294938 Chr.17: 82058358-82058358 on Build GRCh38
rs759872390 Chr.17: 82058309-82058309 on Build GRCh38
rs546981229 Chr.17: 82058335-82058335 on Build GRCh38
rs367654816 Chr.17: 82058279-82058279 on Build GRCh38
rs147637107 Chr.17: 82058387-82058387 on Build GRCh38
rs149720351 Chr.17: 82058231-82058231 on Build GRCh38
rs765728253 Chr.17: 82058264-82058264 on Build GRCh38
rs563712584 Chr.17: 82058293-82058293 on Build GRCh38
rs894630742 Chr.17: 82058238-82058238 on Build GRCh38
rs199715310 Chr.17: 82058289-82058289 on Build GRCh38
rs145545122 Chr.17: 82058250-82058250 on Build GRCh38

View Full Product Details