Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69643140-69643365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND1
Entrez Gene ID
595
Gene Name
cyclin D1
Gene Aliases
BCL1, D11S287E, PRAD1, U21B31
UniGene
Hs.523852
Gene Chromosome Location
Chr.11: 69641105-69654474 on Build GRCh38
Transcripts
NM_053056.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4626899 c.347C>T p.T116I Chr.11: 69643179-69643179 on Build GRCh38
931385 c.318C>T p.C106C Chr.11: 69643150-69643150 on Build GRCh38
6928441 c.385G>A p.D129N Chr.11: 69643217-69643217 on Build GRCh38
6977298 c.336G>C p.K112N Chr.11: 69643168-69643168 on Build GRCh38
6977240 c.339G>A p.M113I Chr.11: 69643171-69643171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755625546 Chr.11: 69643262-69643262 on Build GRCh38
rs955585635 Chr.11: 69643324-69643324 on Build GRCh38
rs746007547 Chr.11: 69643302-69643302 on Build GRCh38
rs752157015 Chr.11: 69643241-69643241 on Build GRCh38
rs367683590 Chr.11: 69643244-69643244 on Build GRCh38
rs1037944680 Chr.11: 69643231-69643231 on Build GRCh38
rs571311003 Chr.11: 69643278-69643278 on Build GRCh38
rs774409065 Chr.11: 69643200-69643200 on Build GRCh38
rs939398424 Chr.11: 69643285-69643285 on Build GRCh38
rs374998781 Chr.11: 69643173-69643173 on Build GRCh38
rs371689281 Chr.11: 69643219-69643219 on Build GRCh38
rs763064872 Chr.11: 69643316-69643316 on Build GRCh38
rs774886294 Chr.11: 69643334-69643334 on Build GRCh38
rs1050971 Chr.11: 69643220-69643220 on Build GRCh38
rs999579613 Chr.11: 69643279-69643279 on Build GRCh38
rs773078314 Chr.11: 69643301-69643301 on Build GRCh38
rs150143978 Chr.11: 69643165-69643165 on Build GRCh38
rs749115370 Chr.11: 69643195-69643195 on Build GRCh38
rs552277149 Chr.11: 69643263-69643264 on Build GRCh38
rs768334579 Chr.11: 69643196-69643196 on Build GRCh38
rs765498502 Chr.11: 69643232-69643232 on Build GRCh38
rs775568711 Chr.11: 69643234-69643234 on Build GRCh38
rs3212864 Chr.11: 69643265-69643265 on Build GRCh38
rs376692814 Chr.11: 69643267-69643267 on Build GRCh38
rs764335132 Chr.11: 69643236-69643236 on Build GRCh38
rs376384298 Chr.11: 69643228-69643228 on Build GRCh38
rs550546811 Chr.11: 69643326-69643326 on Build GRCh38
rs886353624 Chr.11: 69643328-69643328 on Build GRCh38
rs1018607807 Chr.11: 69643207-69643207 on Build GRCh38
rs866931401 Chr.11: 69643230-69643230 on Build GRCh38
rs756875904 Chr.11: 69643162-69643162 on Build GRCh38
rs769760391 Chr.11: 69643307-69643307 on Build GRCh38
rs749956055 Chr.11: 69643260-69643260 on Build GRCh38
rs538688519 Chr.11: 69643309-69643309 on Build GRCh38
rs761932664 Chr.11: 69643290-69643290 on Build GRCh38
rs771952150 Chr.11: 69643217-69643217 on Build GRCh38
rs915462995 Chr.11: 69643266-69643267 on Build GRCh38
rs1055190688 Chr.11: 69643288-69643288 on Build GRCh38
rs768469679 Chr.11: 69643264-69643270 on Build GRCh38
rs759551813 Chr.11: 69643224-69643224 on Build GRCh38
rs1131439 Chr.11: 69643221-69643221 on Build GRCh38
rs748059951 Chr.11: 69643203-69643203 on Build GRCh38
rs747902170 Chr.11: 69643280-69643280 on Build GRCh38
rs1028893771 Chr.11: 69643259-69643260 on Build GRCh38
rs779516743 Chr.11: 69643192-69643192 on Build GRCh38
rs768873833 Chr.11: 69643323-69643323 on Build GRCh38
rs755854763 Chr.11: 69643182-69643182 on Build GRCh38
rs900779300 Chr.11: 69643240-69643240 on Build GRCh38
rs763443319 Chr.11: 69643235-69643235 on Build GRCh38

View Full Product Details