Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118308292-118308551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3E
Entrez Gene ID
916
Gene Name
CD3e molecule
Gene Aliases
IMD18, T3E, TCRE
UniGene
Hs.3003
Gene Chromosome Location
Chr.11: 118304580-118316175 on Build GRCh38
Transcripts
NM_000733.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475071 c.79G>A p.E27K Chr.11: 118308435-118308435 on Build GRCh38
3786985 c.71-3C>T p.? Chr.11: 118308424-118308424 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758377331 Chr.11: 118308468-118308468 on Build GRCh38
rs201812463 Chr.11: 118308491-118308491 on Build GRCh38
rs201307774 Chr.11: 118308405-118308405 on Build GRCh38
rs766052780 Chr.11: 118308472-118308472 on Build GRCh38
rs201478021 Chr.11: 118308458-118308458 on Build GRCh38
rs751100854 Chr.11: 118308473-118308473 on Build GRCh38
rs781468051 Chr.11: 118308404-118308404 on Build GRCh38
rs754653891 Chr.11: 118308402-118308402 on Build GRCh38
rs201867379 Chr.11: 118308429-118308429 on Build GRCh38
rs1056010323 Chr.11: 118308331-118308332 on Build GRCh38
rs34286828 Chr.11: 118308417-118308418 on Build GRCh38
rs77325900 Chr.11: 118308512-118308512 on Build GRCh38
rs377715515 Chr.11: 118308359-118308359 on Build GRCh38
rs957843105 Chr.11: 118308502-118308502 on Build GRCh38
rs1022797779 Chr.11: 118308354-118308354 on Build GRCh38
rs1599810 Chr.11: 118308482-118308482 on Build GRCh38
rs928669985 Chr.11: 118308322-118308322 on Build GRCh38
rs200489032 Chr.11: 118308457-118308457 on Build GRCh38
rs577662207 Chr.11: 118308379-118308379 on Build GRCh38
rs199550655 Chr.11: 118308452-118308452 on Build GRCh38
rs759815699 Chr.11: 118308438-118308438 on Build GRCh38
rs544225142 Chr.11: 118308495-118308495 on Build GRCh38
rs1026969376 Chr.11: 118308397-118308397 on Build GRCh38
rs749862826 Chr.11: 118308493-118308493 on Build GRCh38
rs113849241 Chr.11: 118308387-118308387 on Build GRCh38
rs898653179 Chr.11: 118308353-118308353 on Build GRCh38
rs1021255752 Chr.11: 118308504-118308504 on Build GRCh38
rs757934174 Chr.11: 118308384-118308384 on Build GRCh38
rs908640922 Chr.11: 118308425-118308425 on Build GRCh38
rs371830720 Chr.11: 118308448-118308448 on Build GRCh38
rs749389422 Chr.11: 118308412-118308412 on Build GRCh38
rs375086376 Chr.11: 118308492-118308492 on Build GRCh38
rs201481472 Chr.11: 118308385-118308385 on Build GRCh38
rs750436388 Chr.11: 118308461-118308461 on Build GRCh38
rs1599809 Chr.11: 118308485-118308485 on Build GRCh38
rs568905283 Chr.11: 118308320-118308320 on Build GRCh38
rs114601342 Chr.11: 118308503-118308503 on Build GRCh38
rs969956053 Chr.11: 118308358-118308358 on Build GRCh38
rs200865017 Chr.11: 118308410-118308410 on Build GRCh38
rs770202801 Chr.11: 118308408-118308408 on Build GRCh38
rs372966541 Chr.11: 118308416-118308416 on Build GRCh38
rs771042139 Chr.11: 118308414-118308414 on Build GRCh38
rs537876216 Chr.11: 118308323-118308323 on Build GRCh38
rs17122060 Chr.11: 118308327-118308327 on Build GRCh38

View Full Product Details