Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91273023-91273248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88C
Entrez Gene ID
440193
Gene Name
coiled-coil domain containing 88C
Gene Aliases
DAPLE, HKRP2, KIAA1509, SCA40
UniGene
Hs.525536
Gene Chromosome Location
Chr.14: 91271323-91418498 on Build GRCh38
Transcripts
NM_001080414.3 XM_011536796.2 XM_017021336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5681637 c.1005G>T p.L335L Chr.14: 91273144-91273144 on Build GRCh38
4567097 c.1097_1098CC>TT p.S366F Chr.14: 91273051-91273052 on Build GRCh38
5971287 c.5486T>C p.L1829P Chr.14: 91273226-91273226 on Build GRCh38
5745231 c.935A>G p.E312G Chr.14: 91273214-91273214 on Build GRCh38
6402982 c.1118A>T p.E373V Chr.14: 91273031-91273031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868583429 Chr.14: 91273211-91273211 on Build GRCh38
rs373062767 Chr.14: 91273221-91273221 on Build GRCh38
rs764475037 Chr.14: 91273142-91273142 on Build GRCh38
rs763090813 Chr.14: 91273140-91273140 on Build GRCh38
rs780094181 Chr.14: 91273072-91273072 on Build GRCh38
rs755967067 Chr.14: 91273070-91273070 on Build GRCh38
rs747824457 Chr.14: 91273082-91273082 on Build GRCh38
rs775757498 Chr.14: 91273139-91273139 on Build GRCh38
rs752245028 Chr.14: 91273176-91273176 on Build GRCh38
rs369069440 Chr.14: 91273128-91273128 on Build GRCh38
rs747442301 Chr.14: 91273107-91273107 on Build GRCh38
rs1021776960 Chr.14: 91273164-91273164 on Build GRCh38
rs369450959 Chr.14: 91273218-91273218 on Build GRCh38
rs777000983 Chr.14: 91273050-91273050 on Build GRCh38
rs928474895 Chr.14: 91273069-91273069 on Build GRCh38
rs867368200 Chr.14: 91273094-91273094 on Build GRCh38
rs867416617 Chr.14: 91273178-91273178 on Build GRCh38
rs753850014 Chr.14: 91273165-91273165 on Build GRCh38
rs866896306 Chr.14: 91273181-91273181 on Build GRCh38
rs529377874 Chr.14: 91273088-91273088 on Build GRCh38
rs866438382 Chr.14: 91273179-91273179 on Build GRCh38
rs757901034 Chr.14: 91273193-91273193 on Build GRCh38
rs61742126 Chr.14: 91273058-91273058 on Build GRCh38
rs751814978 Chr.14: 91273147-91273147 on Build GRCh38
rs759673774 Chr.14: 91273051-91273051 on Build GRCh38
rs567110504 Chr.14: 91273077-91273077 on Build GRCh38
rs766949856 Chr.14: 91273044-91273044 on Build GRCh38
rs1007853976 Chr.14: 91273083-91273083 on Build GRCh38
rs777376455 Chr.14: 91273100-91273100 on Build GRCh38
rs200488874 Chr.14: 91273134-91273134 on Build GRCh38
rs764385146 Chr.14: 91273064-91273064 on Build GRCh38
rs771141164 Chr.14: 91273113-91273113 on Build GRCh38
rs746220123 Chr.14: 91273224-91273224 on Build GRCh38
rs374858033 Chr.14: 91273167-91273167 on Build GRCh38
rs537806251 Chr.14: 91273204-91273204 on Build GRCh38
rs374701363 Chr.14: 91273059-91273059 on Build GRCh38
rs754523261 Chr.14: 91273076-91273076 on Build GRCh38
rs766024662 Chr.14: 91273161-91273161 on Build GRCh38
rs760535644 Chr.14: 91273159-91273159 on Build GRCh38
rs565141763 Chr.14: 91273043-91273043 on Build GRCh38
rs568924833 Chr.14: 91273168-91273168 on Build GRCh38
rs749124708 Chr.14: 91273073-91273073 on Build GRCh38
rs1031877266 Chr.14: 91273133-91273133 on Build GRCh38
rs980364075 Chr.14: 91273189-91273189 on Build GRCh38
rs777198297 Chr.14: 91273126-91273126 on Build GRCh38
rs547613648 Chr.14: 91273063-91273063 on Build GRCh38
rs749782727 Chr.14: 91273049-91273050 on Build GRCh38
rs771761723 Chr.14: 91273086-91273086 on Build GRCh38
rs771355988 Chr.14: 91273205-91273205 on Build GRCh38
rs776764078 Chr.14: 91273202-91273202 on Build GRCh38
rs1012188286 Chr.14: 91273156-91273156 on Build GRCh38
rs770239654 Chr.14: 91273226-91273226 on Build GRCh38
rs750278954 Chr.14: 91273065-91273065 on Build GRCh38

View Full Product Details