Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 113003887-113004146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP2
Entrez Gene ID
167227
Gene Name
decapping mRNA 2
Gene Aliases
NUDT20
UniGene
Hs.443875
Gene Chromosome Location
Chr.5: 112976702-113022195 on Build GRCh38
Transcripts
NM_001242377.1 NM_152624.5 NR_038352.1 XM_017009163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5867054 c.841C>T p.P281S Chr.5: 113003976-113003976 on Build GRCh38
7082656 c.859G>T p.D287Y Chr.5: 113003994-113003994 on Build GRCh38
4998208 c.877A>G p.R293G Chr.5: 113004012-113004012 on Build GRCh38
1059678 c.831G>A p.Q277Q Chr.5: 113003966-113003966 on Build GRCh38
6580056 c.835T>G p.L279V Chr.5: 113003970-113003970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142449922 Chr.5: 113004052-113004052 on Build GRCh38
rs748555203 Chr.5: 113003944-113003944 on Build GRCh38
rs1039185646 Chr.5: 113004096-113004096 on Build GRCh38
rs773419110 Chr.5: 113003956-113003956 on Build GRCh38
rs751389450 Chr.5: 113003996-113003996 on Build GRCh38
rs779064426 Chr.5: 113003938-113003938 on Build GRCh38
rs778820671 Chr.5: 113004041-113004041 on Build GRCh38
rs201973039 Chr.5: 113004092-113004092 on Build GRCh38
rs1054082984 Chr.5: 113003964-113003964 on Build GRCh38
rs62364373 Chr.5: 113004114-113004114 on Build GRCh38
rs371078078 Chr.5: 113003969-113003969 on Build GRCh38
rs927184610 Chr.5: 113003950-113003950 on Build GRCh38
rs775093324 Chr.5: 113003972-113003972 on Build GRCh38
rs537890044 Chr.5: 113004003-113004003 on Build GRCh38
rs979969348 Chr.5: 113003935-113003935 on Build GRCh38
rs936772752 Chr.5: 113004055-113004055 on Build GRCh38
rs772667852 Chr.5: 113003945-113003945 on Build GRCh38
rs755274029 Chr.5: 113004039-113004039 on Build GRCh38
rs935909470 Chr.5: 113003955-113003955 on Build GRCh38
rs759200225 Chr.5: 113003966-113003966 on Build GRCh38
rs945761944 Chr.5: 113004012-113004012 on Build GRCh38
rs778408548 Chr.5: 113004060-113004060 on Build GRCh38
rs781555596 Chr.5: 113004093-113004093 on Build GRCh38
rs567645066 Chr.5: 113003973-113003973 on Build GRCh38
rs757188090 Chr.5: 113003997-113003997 on Build GRCh38
rs749378510 Chr.5: 113003920-113003920 on Build GRCh38
rs200344009 Chr.5: 113004035-113004035 on Build GRCh38
rs868675274 Chr.5: 113004004-113004004 on Build GRCh38
rs764138293 Chr.5: 113003992-113003992 on Build GRCh38
rs776200136 Chr.5: 113003963-113003963 on Build GRCh38
rs900724826 Chr.5: 113004105-113004105 on Build GRCh38
rs890919590 Chr.5: 113004086-113004086 on Build GRCh38
rs755045025 Chr.5: 113003921-113003921 on Build GRCh38
rs747430183 Chr.5: 113003957-113003957 on Build GRCh38
rs200919470 Chr.5: 113003981-113003981 on Build GRCh38
rs770564160 Chr.5: 113003960-113003960 on Build GRCh38
rs374218482 Chr.5: 113004058-113004058 on Build GRCh38
rs745478202 Chr.5: 113004103-113004103 on Build GRCh38

View Full Product Details