Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139740946-139741207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11C
Entrez Gene ID
286410
Gene Name
ATPase phospholipid transporting 11C
Gene Aliases
ATPIG, ATPIQ
UniGene
Hs.88252
Gene Chromosome Location
Chr.X: 139726346-139933083 on Build GRCh38
Transcripts
NM_001010986.2 NM_173694.4 XM_017029439.1 XM_017029440.1 XM_017029441.1 XM_017029442.1 XM_017029443.1 XM_017029444.1 XM_017029445.1 XM_017029446.1 XM_017029447.1 XM_017029448.1 XM_017029449.1

SNP Detail

SNP ID Chromosome Location
rs978991958 Chr.X: 139741159-139741159 on Build GRCh38
rs771742509 Chr.X: 139741117-139741117 on Build GRCh38
rs192312820 Chr.X: 139741156-139741156 on Build GRCh38
rs751641983 Chr.X: 139741133-139741133 on Build GRCh38
rs201052330 Chr.X: 139741027-139741027 on Build GRCh38
rs747680720 Chr.X: 139741116-139741116 on Build GRCh38
rs761906009 Chr.X: 139741151-139741151 on Build GRCh38
rs200448452 Chr.X: 139741127-139741127 on Build GRCh38
rs772793430 Chr.X: 139741121-139741121 on Build GRCh38
rs571724082 Chr.X: 139741126-139741126 on Build GRCh38
rs778401112 Chr.X: 139741115-139741115 on Build GRCh38
rs1002019345 Chr.X: 139741123-139741123 on Build GRCh38
rs985403690 Chr.X: 139741103-139741103 on Build GRCh38
rs751761815 Chr.X: 139740980-139740980 on Build GRCh38
rs369906643 Chr.X: 139741124-139741124 on Build GRCh38
rs941171609 Chr.X: 139741100-139741100 on Build GRCh38
rs756474288 Chr.X: 139741078-139741078 on Build GRCh38
rs187463573 Chr.X: 139741132-139741132 on Build GRCh38
rs768098164 Chr.X: 139741106-139741106 on Build GRCh38
rs750778379 Chr.X: 139741069-139741069 on Build GRCh38
rs911175524 Chr.X: 139741110-139741110 on Build GRCh38
rs748860163 Chr.X: 139741099-139741111 on Build GRCh38
rs761844299 Chr.X: 139741146-139741146 on Build GRCh38
rs780497379 Chr.X: 139741101-139741101 on Build GRCh38
rs151005790 Chr.X: 139741062-139741062 on Build GRCh38

View Full Product Details