Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58132909-58133180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
UniGene
Hs.26766
Gene Chromosome Location
Chr.14: 57993536-58152305 on Build GRCh38
Transcripts
NM_001001872.3 NM_001320173.1 XM_011536471.2 XM_017021029.1 XM_017021030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
956493 c.1589C>A p.P530H Chr.14: 58133122-58133122 on Build GRCh38
956494 c.1547G>T p.R516I Chr.14: 58133164-58133164 on Build GRCh38
3956101 c.1621+9G>A p.? Chr.14: 58133081-58133081 on Build GRCh38
3814975 c.1540T>C p.S514P Chr.14: 58133171-58133171 on Build GRCh38
5570090 c.1567A>G p.T523A Chr.14: 58133144-58133144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547933014 Chr.14: 58132988-58132988 on Build GRCh38
rs763531636 Chr.14: 58133152-58133152 on Build GRCh38
rs777413485 Chr.14: 58133056-58133056 on Build GRCh38
rs1020428352 Chr.14: 58132963-58132963 on Build GRCh38
rs776841964 Chr.14: 58133096-58133096 on Build GRCh38
rs1001540275 Chr.14: 58132956-58132957 on Build GRCh38
rs767508866 Chr.14: 58133141-58133141 on Build GRCh38
rs374921811 Chr.14: 58132969-58132972 on Build GRCh38
rs758783966 Chr.14: 58133075-58133075 on Build GRCh38
rs755589596 Chr.14: 58133148-58133148 on Build GRCh38
rs149751032 Chr.14: 58132957-58132958 on Build GRCh38
rs534577596 Chr.14: 58133156-58133156 on Build GRCh38
rs766991382 Chr.14: 58133048-58133048 on Build GRCh38
rs889507220 Chr.14: 58132951-58132951 on Build GRCh38
rs960664407 Chr.14: 58133007-58133007 on Build GRCh38
rs1045807665 Chr.14: 58133100-58133100 on Build GRCh38
rs755716909 Chr.14: 58133054-58133054 on Build GRCh38
rs920496818 Chr.14: 58133010-58133010 on Build GRCh38
rs748902828 Chr.14: 58133059-58133059 on Build GRCh38
rs370200646 Chr.14: 58133126-58133126 on Build GRCh38
rs12886921 Chr.14: 58133129-58133129 on Build GRCh38
rs747490700 Chr.14: 58133092-58133092 on Build GRCh38
rs759455728 Chr.14: 58133132-58133132 on Build GRCh38
rs536638825 Chr.14: 58133006-58133006 on Build GRCh38
rs1018650551 Chr.14: 58132984-58132984 on Build GRCh38
rs112976465 Chr.14: 58133064-58133064 on Build GRCh38
rs779516583 Chr.14: 58132958-58132959 on Build GRCh38
rs762930514 Chr.14: 58132969-58132969 on Build GRCh38
rs200713244 Chr.14: 58133127-58133127 on Build GRCh38
rs1022026567 Chr.14: 58132986-58132986 on Build GRCh38
rs752308200 Chr.14: 58133049-58133049 on Build GRCh38
rs773427478 Chr.14: 58133124-58133124 on Build GRCh38
rs983794792 Chr.14: 58133038-58133038 on Build GRCh38
rs200874016 Chr.14: 58133147-58133147 on Build GRCh38
rs747341611 Chr.14: 58133054-58133054 on Build GRCh38
rs769768431 Chr.14: 58133123-58133123 on Build GRCh38
rs780733063 Chr.14: 58133050-58133051 on Build GRCh38
rs377061784 Chr.14: 58133103-58133103 on Build GRCh38
rs992546439 Chr.14: 58133036-58133036 on Build GRCh38
rs184011903 Chr.14: 58132989-58132989 on Build GRCh38
rs967837073 Chr.14: 58132987-58132987 on Build GRCh38
rs987146600 Chr.14: 58132975-58132975 on Build GRCh38
rs1010808674 Chr.14: 58132957-58132960 on Build GRCh38
rs916598602 Chr.14: 58133057-58133057 on Build GRCh38
rs779540957 Chr.14: 58133005-58133005 on Build GRCh38
rs905830751 Chr.14: 58132940-58132940 on Build GRCh38
rs780594136 Chr.14: 58133081-58133081 on Build GRCh38
rs772401630 Chr.14: 58133045-58133046 on Build GRCh38
rs372947247 Chr.14: 58133130-58133130 on Build GRCh38
rs552079842 Chr.14: 58133093-58133093 on Build GRCh38
rs199910312 Chr.14: 58133136-58133136 on Build GRCh38

View Full Product Details