Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101956534-101956785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
-
UniGene
Hs.16004
Gene Chromosome Location
Chr.10: 101845599-102056237 on Build GRCh38
Transcripts
NM_024541.2 XM_006717969.1 XM_011540151.2 XM_011540152.2 XM_011540153.2 XM_011540154.2 XM_011540156.2 XM_011540157.2 XM_017016635.1 XM_017016636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786307 c.1666A>G p.I556V Chr.10: 101956636-101956636 on Build GRCh38
141310 c.1610A>T p.Y537F Chr.10: 101956692-101956692 on Build GRCh38
4011168 c.1614C>T p.G538G Chr.10: 101956688-101956688 on Build GRCh38
5441483 c.1695C>G p.L565L Chr.10: 101956607-101956607 on Build GRCh38
5631692 c.1612G>A p.G538S Chr.10: 101956690-101956690 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375360848 Chr.10: 101956574-101956574 on Build GRCh38
rs771732721 Chr.10: 101956685-101956685 on Build GRCh38
rs759389501 Chr.10: 101956722-101956722 on Build GRCh38
rs752518171 Chr.10: 101956582-101956582 on Build GRCh38
rs778284189 Chr.10: 101956752-101956752 on Build GRCh38
rs368670935 Chr.10: 101956742-101956742 on Build GRCh38
rs369660890 Chr.10: 101956611-101956611 on Build GRCh38
rs570122508 Chr.10: 101956723-101956723 on Build GRCh38
rs537178610 Chr.10: 101956745-101956745 on Build GRCh38
rs766126657 Chr.10: 101956689-101956689 on Build GRCh38
rs994151618 Chr.10: 101956562-101956562 on Build GRCh38
rs758509939 Chr.10: 101956605-101956605 on Build GRCh38
rs772828942 Chr.10: 101956687-101956687 on Build GRCh38
rs1026958941 Chr.10: 101956587-101956587 on Build GRCh38
rs1006936774 Chr.10: 101956712-101956712 on Build GRCh38
rs749723246 Chr.10: 101956671-101956671 on Build GRCh38
rs372697789 Chr.10: 101956746-101956746 on Build GRCh38
rs757446410 Chr.10: 101956624-101956624 on Build GRCh38
rs934088600 Chr.10: 101956647-101956647 on Build GRCh38
rs888956931 Chr.10: 101956674-101956674 on Build GRCh38
rs539893616 Chr.10: 101956704-101956704 on Build GRCh38
rs781574136 Chr.10: 101956632-101956632 on Build GRCh38
rs372532953 Chr.10: 101956666-101956666 on Build GRCh38
rs367986699 Chr.10: 101956664-101956664 on Build GRCh38
rs752710792 Chr.10: 101956737-101956737 on Build GRCh38
rs375031309 Chr.10: 101956688-101956688 on Build GRCh38
rs746236094 Chr.10: 101956651-101956651 on Build GRCh38
rs965279251 Chr.10: 101956753-101956753 on Build GRCh38
rs776347179 Chr.10: 101956699-101956699 on Build GRCh38

View Full Product Details