Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76175496-76175727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929439
Entrez Gene ID
101929439
Gene Name
uncharacterized LOC101929439
Gene Aliases
-
UniGene
Hs.631721
Gene Chromosome Location
Chr.15: 76174891-76181486 on Build GRCh38
Transcripts
NR_120360.1 NM_152335.2 XM_005254160.2 XM_017021915.1 XM_017021916.1 XM_017021917.1

Gene Symbol
TMEM266
Entrez Gene ID
123591
Gene Name
transmembrane protein 266
Gene Aliases
C15orf27, HVRP1
UniGene
Hs.661411
Gene Chromosome Location
Chr.15: 76059929-76204963 on Build GRCh38
Transcripts
NR_120360.1 NM_152335.2 XM_005254160.2 XM_017021915.1 XM_017021916.1 XM_017021917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1939743 c.746C>T p.T249M Chr.15: 76175652-76175652 on Build GRCh38
1374658 c.673C>T p.L225L Chr.15: 76175579-76175579 on Build GRCh38
1374659 c.702C>T p.Y234Y Chr.15: 76175608-76175608 on Build GRCh38
5509619 c.658G>A p.V220I Chr.15: 76175564-76175564 on Build GRCh38
342198 c.734T>C p.L245P Chr.15: 76175640-76175640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897889853 Chr.15: 76175674-76175674 on Build GRCh38
rs200737085 Chr.15: 76175563-76175563 on Build GRCh38
rs181008068 Chr.15: 76175684-76175684 on Build GRCh38
rs942023218 Chr.15: 76175630-76175630 on Build GRCh38
rs957993029 Chr.15: 76175523-76175523 on Build GRCh38
rs778038275 Chr.15: 76175624-76175624 on Build GRCh38
rs772544145 Chr.15: 76175652-76175652 on Build GRCh38
rs200150323 Chr.15: 76175566-76175566 on Build GRCh38
rs753283518 Chr.15: 76175688-76175688 on Build GRCh38
rs774799142 Chr.15: 76175673-76175673 on Build GRCh38
rs201768606 Chr.15: 76175572-76175572 on Build GRCh38
rs771311818 Chr.15: 76175637-76175637 on Build GRCh38
rs777395226 Chr.15: 76175704-76175704 on Build GRCh38
rs752520367 Chr.15: 76175621-76175621 on Build GRCh38
rs752614030 Chr.15: 76175694-76175694 on Build GRCh38
rs373335165 Chr.15: 76175564-76175564 on Build GRCh38
rs778726773 Chr.15: 76175614-76175614 on Build GRCh38
rs375786494 Chr.15: 76175579-76175579 on Build GRCh38
rs773359873 Chr.15: 76175519-76175519 on Build GRCh38
rs775930403 Chr.15: 76175653-76175653 on Build GRCh38
rs535708773 Chr.15: 76175692-76175692 on Build GRCh38
rs763347104 Chr.15: 76175656-76175656 on Build GRCh38
rs1000921179 Chr.15: 76175707-76175707 on Build GRCh38
rs771188829 Chr.15: 76175543-76175543 on Build GRCh38
rs368931639 Chr.15: 76175644-76175644 on Build GRCh38
rs765824878 Chr.15: 76175608-76175608 on Build GRCh38
rs760082559 Chr.15: 76175562-76175562 on Build GRCh38
rs765214179 Chr.15: 76175693-76175693 on Build GRCh38
rs781358139 Chr.15: 76175639-76175639 on Build GRCh38
rs376566174 Chr.15: 76175687-76175687 on Build GRCh38
rs557513678 Chr.15: 76175703-76175703 on Build GRCh38
rs974806716 Chr.15: 76175539-76175539 on Build GRCh38
rs749641078 Chr.15: 76175545-76175546 on Build GRCh38
rs750081808 Chr.15: 76175582-76175582 on Build GRCh38
rs747155045 Chr.15: 76175632-76175632 on Build GRCh38
rs760155139 Chr.15: 76175594-76175594 on Build GRCh38
rs1057295429 Chr.15: 76175690-76175690 on Build GRCh38
rs761300568 Chr.15: 76175609-76175609 on Build GRCh38
rs758190261 Chr.15: 76175623-76175623 on Build GRCh38
rs772277688 Chr.15: 76175516-76175516 on Build GRCh38
rs912498287 Chr.15: 76175648-76175648 on Build GRCh38
rs776875057 Chr.15: 76175553-76175553 on Build GRCh38
rs373481496 Chr.15: 76175669-76175669 on Build GRCh38
rs769321159 Chr.15: 76175633-76175633 on Build GRCh38
rs201795006 Chr.15: 76175612-76175612 on Build GRCh38
rs747084863 Chr.15: 76175532-76175532 on Build GRCh38

View Full Product Details