Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70452353-70452592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135A
Entrez Gene ID
57579
Gene Name
family with sequence similarity 135 member A
Gene Aliases
KIAA1411
UniGene
Hs.211700
Gene Chromosome Location
Chr.6: 70413404-70561174 on Build GRCh38
Transcripts
NM_001105531.2 NM_001162529.1 NM_020819.4 XM_005248743.1 XM_005248744.4 XM_005248745.2 XM_005248748.2 XM_005248749.1 XM_005248751.2 XM_011535993.2 XM_011535994.2 XM_011535995.2 XM_011535996.2 XM_011535997.2 XM_017011124.1 XM_017011125.1 XM_017011126.1 XM_017011127.1 XM_017011128.1 XM_017011129.1 XM_017011130.1 XM_017011131.1 XM_017011132.1 XM_017011133.1 XM_017011134.1 XM_017011135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445694 c.78-2A>C p.? Chr.6: 70452490-70452490 on Build GRCh38
1496446 c.113C>G p.S38* Chr.6: 70452527-70452527 on Build GRCh38
4766370 c.91C>T p.R31C Chr.6: 70452505-70452505 on Build GRCh38
3875222 c.142T>A p.L48M Chr.6: 70452556-70452556 on Build GRCh38
743241 c.86A>G p.Q29R Chr.6: 70452500-70452500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746791952 Chr.6: 70452542-70452542 on Build GRCh38
rs781287498 Chr.6: 70452562-70452562 on Build GRCh38
rs748185218 Chr.6: 70452484-70452484 on Build GRCh38
rs764762660 Chr.6: 70452523-70452523 on Build GRCh38
rs568674023 Chr.6: 70452433-70452433 on Build GRCh38
rs551119539 Chr.6: 70452505-70452505 on Build GRCh38
rs747280320 Chr.6: 70452469-70452469 on Build GRCh38
rs187205603 Chr.6: 70452399-70452399 on Build GRCh38
rs753471313 Chr.6: 70452514-70452514 on Build GRCh38
rs375692237 Chr.6: 70452464-70452464 on Build GRCh38
rs537609415 Chr.6: 70452456-70452456 on Build GRCh38
rs773617245 Chr.6: 70452502-70452502 on Build GRCh38
rs778728718 Chr.6: 70452459-70452459 on Build GRCh38
rs370276504 Chr.6: 70452476-70452476 on Build GRCh38
rs3736752 Chr.6: 70452427-70452427 on Build GRCh38
rs754757798 Chr.6: 70452550-70452550 on Build GRCh38
rs1038981284 Chr.6: 70452501-70452501 on Build GRCh38
rs768982914 Chr.6: 70452477-70452477 on Build GRCh38
rs763659151 Chr.6: 70452510-70452510 on Build GRCh38
rs898607337 Chr.6: 70452444-70452444 on Build GRCh38
rs1038358946 Chr.6: 70452435-70452435 on Build GRCh38
rs758920697 Chr.6: 70452432-70452432 on Build GRCh38
rs201194449 Chr.6: 70452508-70452508 on Build GRCh38
rs373906503 Chr.6: 70452537-70452537 on Build GRCh38
rs758384489 Chr.6: 70452529-70452529 on Build GRCh38
rs779961624 Chr.6: 70452466-70452466 on Build GRCh38
rs539850700 Chr.6: 70452528-70452528 on Build GRCh38
rs764399547 Chr.6: 70452551-70452551 on Build GRCh38
rs147103878 Chr.6: 70452506-70452506 on Build GRCh38
rs760379557 Chr.6: 70452509-70452509 on Build GRCh38
rs745768221 Chr.6: 70452462-70452462 on Build GRCh38
rs776840875 Chr.6: 70452480-70452480 on Build GRCh38
rs756739363 Chr.6: 70452522-70452522 on Build GRCh38
rs769857766 Chr.6: 70452490-70452490 on Build GRCh38

View Full Product Details