Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45050586-45050837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBX2
Entrez Gene ID
440097
Gene Name
developing brain homeobox 2
Gene Aliases
-
UniGene
Hs.302764
Gene Chromosome Location
Chr.12: 45014756-45051099 on Build GRCh38
Transcripts
NM_001004329.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4632036 c.301A>G p.T101A Chr.12: 45050627-45050627 on Build GRCh38
6049254 c.302C>T p.T101M Chr.12: 45050626-45050626 on Build GRCh38
5667415 c.339C>G p.D113E Chr.12: 45050589-45050589 on Build GRCh38
4676308 c.153G>A p.T51T Chr.12: 45050775-45050775 on Build GRCh38
227100 c.142G>A p.G48S Chr.12: 45050786-45050786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377646614 Chr.12: 45050623-45050623 on Build GRCh38
rs750311548 Chr.12: 45050748-45050748 on Build GRCh38
rs750113512 Chr.12: 45050629-45050629 on Build GRCh38
rs979408201 Chr.12: 45050768-45050768 on Build GRCh38
rs779078501 Chr.12: 45050818-45050819 on Build GRCh38
rs573108342 Chr.12: 45050793-45050793 on Build GRCh38
rs139910495 Chr.12: 45050733-45050733 on Build GRCh38
rs754491176 Chr.12: 45050687-45050687 on Build GRCh38
rs1055830362 Chr.12: 45050643-45050643 on Build GRCh38
rs1048740449 Chr.12: 45050717-45050717 on Build GRCh38
rs929775800 Chr.12: 45050624-45050624 on Build GRCh38
rs760536409 Chr.12: 45050756-45050756 on Build GRCh38
rs774894428 Chr.12: 45050627-45050627 on Build GRCh38
rs753499951 Chr.12: 45050766-45050766 on Build GRCh38
rs776898064 Chr.12: 45050725-45050725 on Build GRCh38
rs534192224 Chr.12: 45050808-45050808 on Build GRCh38
rs748646032 Chr.12: 45050700-45050700 on Build GRCh38
rs767910557 Chr.12: 45050664-45050664 on Build GRCh38
rs911327328 Chr.12: 45050737-45050737 on Build GRCh38
rs779332087 Chr.12: 45050697-45050697 on Build GRCh38
rs894150392 Chr.12: 45050646-45050646 on Build GRCh38
rs968296283 Chr.12: 45050681-45050681 on Build GRCh38
rs754475024 Chr.12: 45050780-45050780 on Build GRCh38
rs994502797 Chr.12: 45050708-45050708 on Build GRCh38
rs778049675 Chr.12: 45050709-45050709 on Build GRCh38
rs200382293 Chr.12: 45050648-45050648 on Build GRCh38
rs926680426 Chr.12: 45050631-45050631 on Build GRCh38
rs767425325 Chr.12: 45050744-45050744 on Build GRCh38
rs571961238 Chr.12: 45050622-45050622 on Build GRCh38
rs1020862088 Chr.12: 45050753-45050753 on Build GRCh38
rs1046908271 Chr.12: 45050775-45050775 on Build GRCh38
rs1018655526 Chr.12: 45050743-45050743 on Build GRCh38
rs759582033 Chr.12: 45050728-45050728 on Build GRCh38
rs955602267 Chr.12: 45050722-45050722 on Build GRCh38
rs919826931 Chr.12: 45050746-45050746 on Build GRCh38
rs761785803 Chr.12: 45050740-45050740 on Build GRCh38
rs758832490 Chr.12: 45050799-45050799 on Build GRCh38
rs999637600 Chr.12: 45050696-45050696 on Build GRCh38
rs913808686 Chr.12: 45050612-45050612 on Build GRCh38
rs778435010 Chr.12: 45050789-45050789 on Build GRCh38
rs771208101 Chr.12: 45050720-45050720 on Build GRCh38
rs373749684 Chr.12: 45050617-45050617 on Build GRCh38
rs200605636 Chr.12: 45050636-45050636 on Build GRCh38
rs772445960 Chr.12: 45050702-45050702 on Build GRCh38
rs536313056 Chr.12: 45050719-45050719 on Build GRCh38
rs766187055 Chr.12: 45050759-45050759 on Build GRCh38
rs774179347 Chr.12: 45050739-45050739 on Build GRCh38
rs940187028 Chr.12: 45050684-45050684 on Build GRCh38
rs1037283461 Chr.12: 45050691-45050691 on Build GRCh38
rs569493933 Chr.12: 45050698-45050698 on Build GRCh38
rs898779472 Chr.12: 45050701-45050701 on Build GRCh38

View Full Product Details