Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 28507541-28507812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB5
Entrez Gene ID
9586
Gene Name
cAMP responsive element binding protein 5
Gene Aliases
CRE-BPA, CREB-5
UniGene
Hs.437075
Gene Chromosome Location
Chr.7: 28299321-28825894 on Build GRCh38
Transcripts
NM_004904.3 NM_182898.3 NM_182899.4 XM_005249906.1 XM_017012806.1 XM_017012807.1 XM_017012808.1 XM_017012809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7236555 c.234G>A p.L78L Chr.7: 28507680-28507680 on Build GRCh38
5710531 c.290G>A p.R97Q Chr.7: 28507736-28507736 on Build GRCh38
5762547 c.218G>T p.G73V Chr.7: 28507664-28507664 on Build GRCh38
7362632 c.254A>T p.E85V Chr.7: 28507700-28507700 on Build GRCh38
3832600 c.291G>T p.R97R Chr.7: 28507737-28507737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766189534 Chr.7: 28507735-28507735 on Build GRCh38
rs575115266 Chr.7: 28507715-28507715 on Build GRCh38
rs763500648 Chr.7: 28507708-28507708 on Build GRCh38
rs770302370 Chr.7: 28507697-28507697 on Build GRCh38
rs767456309 Chr.7: 28507635-28507635 on Build GRCh38
rs781117430 Chr.7: 28507581-28507581 on Build GRCh38
rs374079519 Chr.7: 28507789-28507789 on Build GRCh38
rs762102181 Chr.7: 28507634-28507634 on Build GRCh38
rs747555411 Chr.7: 28507698-28507698 on Build GRCh38
rs374506501 Chr.7: 28507785-28507785 on Build GRCh38
rs937186996 Chr.7: 28507654-28507654 on Build GRCh38
rs747834326 Chr.7: 28507602-28507602 on Build GRCh38
rs764383622 Chr.7: 28507633-28507633 on Build GRCh38
rs750199128 Chr.7: 28507636-28507636 on Build GRCh38
rs991847862 Chr.7: 28507674-28507674 on Build GRCh38
rs370655980 Chr.7: 28507593-28507593 on Build GRCh38
rs778473920 Chr.7: 28507668-28507668 on Build GRCh38
rs756142208 Chr.7: 28507637-28507637 on Build GRCh38
rs371067938 Chr.7: 28507758-28507758 on Build GRCh38
rs780441733 Chr.7: 28507775-28507775 on Build GRCh38
rs746185795 Chr.7: 28507696-28507696 on Build GRCh38
rs772126660 Chr.7: 28507731-28507731 on Build GRCh38
rs745846419 Chr.7: 28507586-28507586 on Build GRCh38
rs893051792 Chr.7: 28507607-28507607 on Build GRCh38
rs771682117 Chr.7: 28507680-28507680 on Build GRCh38
rs760077074 Chr.7: 28507625-28507625 on Build GRCh38
rs767799448 Chr.7: 28507722-28507722 on Build GRCh38
rs764327382 Chr.7: 28507575-28507576 on Build GRCh38
rs781572047 Chr.7: 28507695-28507695 on Build GRCh38
rs768366822 Chr.7: 28507598-28507598 on Build GRCh38
rs752211944 Chr.7: 28507747-28507747 on Build GRCh38
rs746238949 Chr.7: 28507759-28507759 on Build GRCh38
rs754514486 Chr.7: 28507660-28507660 on Build GRCh38
rs756082984 Chr.7: 28507590-28507590 on Build GRCh38
rs201457566 Chr.7: 28507757-28507757 on Build GRCh38
rs780080880 Chr.7: 28507641-28507641 on Build GRCh38
rs200202352 Chr.7: 28507672-28507672 on Build GRCh38
rs768663791 Chr.7: 28507711-28507711 on Build GRCh38
rs145890705 Chr.7: 28507620-28507620 on Build GRCh38
rs142741982 Chr.7: 28507736-28507736 on Build GRCh38
rs201301576 Chr.7: 28507592-28507592 on Build GRCh38
rs749796951 Chr.7: 28507776-28507776 on Build GRCh38
rs1029667113 Chr.7: 28507790-28507790 on Build GRCh38
rs138761343 Chr.7: 28507643-28507643 on Build GRCh38
rs561442711 Chr.7: 28507784-28507784 on Build GRCh38
rs550409575 Chr.7: 28507677-28507677 on Build GRCh38
rs765698094 Chr.7: 28507626-28507626 on Build GRCh38
rs775664508 Chr.7: 28507629-28507629 on Build GRCh38
rs773807186 Chr.7: 28507601-28507601 on Build GRCh38
rs981535574 Chr.7: 28507632-28507632 on Build GRCh38
rs770212651 Chr.7: 28507763-28507763 on Build GRCh38
rs761926508 Chr.7: 28507787-28507787 on Build GRCh38
rs760478767 Chr.7: 28507732-28507732 on Build GRCh38
rs544934809 Chr.7: 28507650-28507650 on Build GRCh38
rs772426759 Chr.7: 28507788-28507788 on Build GRCh38

View Full Product Details