Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15666042-15666281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC171
Entrez Gene ID
203238
Gene Name
coiled-coil domain containing 171
Gene Aliases
C9orf93, bA536D16.1, bA778P13.1
UniGene
Hs.17267
Gene Chromosome Location
Chr.9: 15552874-16062431 on Build GRCh38
Transcripts
NM_173550.2 XM_005251397.2 XM_005251398.3 XM_005251399.4 XM_011517788.2 XM_011517791.2 XM_011517796.2 XM_017014431.1 XM_017014432.1 XM_017014433.1 XM_017014434.1 XM_017014435.1 XM_017014436.1 XM_017014437.1 XM_017014438.1 XM_017014439.1 XM_017014440.1 XM_017014441.1 XM_017014442.1 XM_017014443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199120 c.1033C>T p.R345* Chr.9: 15666280-15666280 on Build GRCh38
5027051 c.957G>A p.K319K Chr.9: 15666204-15666204 on Build GRCh38
7359883 c.997A>C p.K333Q Chr.9: 15666244-15666244 on Build GRCh38
1461551 c.920G>A p.R307Q Chr.9: 15666167-15666167 on Build GRCh38
5524337 c.925C>T p.R309* Chr.9: 15666172-15666172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200082488 Chr.9: 15666154-15666154 on Build GRCh38
rs773194320 Chr.9: 15666208-15666208 on Build GRCh38
rs200110488 Chr.9: 15666152-15666152 on Build GRCh38
rs202229183 Chr.9: 15666158-15666158 on Build GRCh38
rs764517961 Chr.9: 15666161-15666161 on Build GRCh38
rs773036111 Chr.9: 15666156-15666156 on Build GRCh38
rs921911022 Chr.9: 15666167-15666167 on Build GRCh38
rs201714618 Chr.9: 15666153-15666153 on Build GRCh38
rs780050635 Chr.9: 15666177-15666177 on Build GRCh38
rs769083141 Chr.9: 15666232-15666232 on Build GRCh38
rs530744027 Chr.9: 15666172-15666172 on Build GRCh38
rs769770564 Chr.9: 15666144-15666144 on Build GRCh38
rs751929117 Chr.9: 15666162-15666162 on Build GRCh38
rs200852139 Chr.9: 15666159-15666159 on Build GRCh38
rs370829802 Chr.9: 15666242-15666242 on Build GRCh38
rs770448682 Chr.9: 15666224-15666224 on Build GRCh38
rs965707120 Chr.9: 15666141-15666141 on Build GRCh38
rs138513099 Chr.9: 15666205-15666205 on Build GRCh38
rs749535194 Chr.9: 15666149-15666149 on Build GRCh38
rs887900660 Chr.9: 15666248-15666248 on Build GRCh38
rs1033993654 Chr.9: 15666077-15666077 on Build GRCh38
rs551724590 Chr.9: 15666157-15666157 on Build GRCh38
rs372889968 Chr.9: 15666195-15666195 on Build GRCh38
rs775780876 Chr.9: 15666147-15666147 on Build GRCh38
rs533218926 Chr.9: 15666108-15666108 on Build GRCh38
rs958387462 Chr.9: 15666103-15666103 on Build GRCh38
rs779130651 Chr.9: 15666198-15666198 on Build GRCh38
rs759146540 Chr.9: 15666243-15666243 on Build GRCh38
rs768418835 Chr.9: 15666151-15666151 on Build GRCh38
rs989901021 Chr.9: 15666110-15666110 on Build GRCh38
rs750961495 Chr.9: 15666169-15666169 on Build GRCh38
rs914338347 Chr.9: 15666116-15666116 on Build GRCh38
rs747881762 Chr.9: 15666203-15666203 on Build GRCh38
rs760587640 Chr.9: 15666211-15666211 on Build GRCh38
rs1057209618 Chr.9: 15666173-15666173 on Build GRCh38
rs757715493 Chr.9: 15666166-15666166 on Build GRCh38
rs756209911 Chr.9: 15666175-15666175 on Build GRCh38
rs550127556 Chr.9: 15666122-15666122 on Build GRCh38
rs747492779 Chr.9: 15666115-15666115 on Build GRCh38
rs142027816 Chr.9: 15666178-15666178 on Build GRCh38
rs972672364 Chr.9: 15666146-15666146 on Build GRCh38
rs757876954 Chr.9: 15666120-15666120 on Build GRCh38

View Full Product Details