Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45533676-45533949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
UniGene
Hs.24601
Gene Chromosome Location
Chr.22: 45502839-45601134 on Build GRCh38
Transcripts
NM_001996.3 NM_006485.3 NM_006486.2 NM_006487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6661489 c.731G>A p.R244Q Chr.22: 45533845-45533845 on Build GRCh38
5881271 c.672_673insT p.H225fs*29 Chr.22: 45533786-45533787 on Build GRCh38
6661488 c.731G>A p.R244Q Chr.22: 45533845-45533845 on Build GRCh38
5881270 c.672_673insT p.H225fs*29 Chr.22: 45533786-45533787 on Build GRCh38
6661487 c.731G>A p.R244Q Chr.22: 45533845-45533845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199991452 Chr.22: 45533738-45533738 on Build GRCh38
rs3216851 Chr.22: 45533715-45533715 on Build GRCh38
rs776907746 Chr.22: 45533724-45533724 on Build GRCh38
rs369567067 Chr.22: 45533719-45533719 on Build GRCh38
rs775408699 Chr.22: 45533797-45533797 on Build GRCh38
rs760570023 Chr.22: 45533800-45533800 on Build GRCh38
rs532883593 Chr.22: 45533744-45533744 on Build GRCh38
rs374402590 Chr.22: 45533908-45533908 on Build GRCh38
rs562978224 Chr.22: 45533801-45533801 on Build GRCh38
rs772506222 Chr.22: 45533718-45533718 on Build GRCh38
rs748672066 Chr.22: 45533845-45533845 on Build GRCh38
rs545434511 Chr.22: 45533749-45533749 on Build GRCh38
rs369112052 Chr.22: 45533779-45533779 on Build GRCh38
rs113737102 Chr.22: 45533899-45533899 on Build GRCh38
rs768455798 Chr.22: 45533907-45533907 on Build GRCh38
rs756238855 Chr.22: 45533910-45533920 on Build GRCh38
rs780533565 Chr.22: 45533768-45533768 on Build GRCh38
rs750101464 Chr.22: 45533763-45533763 on Build GRCh38
rs144129499 Chr.22: 45533771-45533771 on Build GRCh38
rs763577133 Chr.22: 45533739-45533739 on Build GRCh38
rs1025624953 Chr.22: 45533862-45533862 on Build GRCh38
rs140981949 Chr.22: 45533814-45533814 on Build GRCh38
rs370004105 Chr.22: 45533910-45533910 on Build GRCh38
rs774464064 Chr.22: 45533887-45533887 on Build GRCh38
rs549459139 Chr.22: 45533844-45533844 on Build GRCh38
rs371726470 Chr.22: 45533780-45533780 on Build GRCh38
rs762096216 Chr.22: 45533731-45533731 on Build GRCh38
rs756260694 Chr.22: 45533850-45533850 on Build GRCh38
rs146491057 Chr.22: 45533775-45533775 on Build GRCh38
rs776568892 Chr.22: 45533802-45533802 on Build GRCh38
rs16994211 Chr.22: 45533734-45533734 on Build GRCh38
rs761252663 Chr.22: 45533809-45533809 on Build GRCh38
rs911021376 Chr.22: 45533914-45533914 on Build GRCh38
rs201033702 Chr.22: 45533722-45533722 on Build GRCh38
rs566373261 Chr.22: 45533915-45533915 on Build GRCh38
rs767352348 Chr.22: 45533736-45533736 on Build GRCh38
rs952609235 Chr.22: 45533896-45533896 on Build GRCh38
rs105199 Chr.22: 45533697-45533697 on Build GRCh38
rs749594164 Chr.22: 45533789-45533789 on Build GRCh38
rs113516927 Chr.22: 45533858-45533858 on Build GRCh38
rs766630351 Chr.22: 45533830-45533830 on Build GRCh38
rs564562355 Chr.22: 45533745-45533745 on Build GRCh38
rs746176353 Chr.22: 45533717-45533717 on Build GRCh38
rs758621512 Chr.22: 45533767-45533767 on Build GRCh38
rs751912120 Chr.22: 45533835-45533835 on Build GRCh38
rs746761923 Chr.22: 45533893-45533893 on Build GRCh38
rs372048171 Chr.22: 45533720-45533720 on Build GRCh38
rs377281271 Chr.22: 45533909-45533909 on Build GRCh38
rs755416957 Chr.22: 45533836-45533836 on Build GRCh38
rs892038183 Chr.22: 45533748-45533748 on Build GRCh38
rs777970776 Chr.22: 45533852-45533852 on Build GRCh38
rs58965835 Chr.22: 45533805-45533805 on Build GRCh38
rs972747723 Chr.22: 45533849-45533849 on Build GRCh38
rs771229284 Chr.22: 45533869-45533869 on Build GRCh38
rs758127225 Chr.22: 45533816-45533816 on Build GRCh38
rs1001988378 Chr.22: 45533834-45533834 on Build GRCh38
rs933597293 Chr.22: 45533791-45533791 on Build GRCh38
rs542978755 Chr.22: 45533796-45533796 on Build GRCh38

View Full Product Details