Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104995663-104995924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP5
Entrez Gene ID
838
Gene Name
caspase 5
Gene Aliases
ICE(rel)III, ICEREL-III, ICH-3
UniGene
Hs.213327
Gene Chromosome Location
Chr.11: 104994240-105023902 on Build GRCh38
Transcripts
NM_001136109.1 NM_001136110.1 NM_001136112.1 NM_004347.3 NR_024239.1 NR_036562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5963053 c.1343G>C p.*448S Chr.11: 104995745-104995745 on Build GRCh38
922431 c.1169C>A p.S390* Chr.11: 104995832-104995832 on Build GRCh38
5590058 c.1166A>T p.K389I Chr.11: 104995835-104995835 on Build GRCh38
3442853 c.1250G>A p.G417D Chr.11: 104995751-104995751 on Build GRCh38
6290884 c.1224G>A p.L408L Chr.11: 104995777-104995777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753549463 Chr.11: 104995698-104995698 on Build GRCh38
rs45478501 Chr.11: 104995881-104995881 on Build GRCh38
rs141361242 Chr.11: 104995788-104995788 on Build GRCh38
rs778639041 Chr.11: 104995717-104995717 on Build GRCh38
rs961686768 Chr.11: 104995897-104995897 on Build GRCh38
rs764586508 Chr.11: 104995879-104995879 on Build GRCh38
rs759285379 Chr.11: 104995719-104995719 on Build GRCh38
rs901698533 Chr.11: 104995807-104995807 on Build GRCh38
rs767099412 Chr.11: 104995831-104995831 on Build GRCh38
rs771379291 Chr.11: 104995758-104995758 on Build GRCh38
rs761410953 Chr.11: 104995783-104995783 on Build GRCh38
rs1013501056 Chr.11: 104995882-104995882 on Build GRCh38
rs779767143 Chr.11: 104995794-104995794 on Build GRCh38
rs138897099 Chr.11: 104995780-104995780 on Build GRCh38
rs560945010 Chr.11: 104995785-104995785 on Build GRCh38
rs763626381 Chr.11: 104995697-104995697 on Build GRCh38
rs748389801 Chr.11: 104995735-104995735 on Build GRCh38
rs763750862 Chr.11: 104995775-104995775 on Build GRCh38
rs778085364 Chr.11: 104995833-104995833 on Build GRCh38
rs748239649 Chr.11: 104995826-104995826 on Build GRCh38
rs749400550 Chr.11: 104995837-104995837 on Build GRCh38
rs368312007 Chr.11: 104995725-104995725 on Build GRCh38
rs772932514 Chr.11: 104995880-104995880 on Build GRCh38
rs3181333 Chr.11: 104995690-104995690 on Build GRCh38
rs373084525 Chr.11: 104995699-104995699 on Build GRCh38
rs372804237 Chr.11: 104995787-104995787 on Build GRCh38
rs760059673 Chr.11: 104995772-104995772 on Build GRCh38
rs760289089 Chr.11: 104995693-104995693 on Build GRCh38
rs201531535 Chr.11: 104995855-104995855 on Build GRCh38
rs199914340 Chr.11: 104995846-104995846 on Build GRCh38
rs1050335756 Chr.11: 104995774-104995774 on Build GRCh38
rs770533387 Chr.11: 104995691-104995691 on Build GRCh38
rs1019707283 Chr.11: 104995791-104995791 on Build GRCh38
rs200552536 Chr.11: 104995902-104995902 on Build GRCh38
rs746884574 Chr.11: 104995728-104995728 on Build GRCh38
rs549693625 Chr.11: 104995832-104995832 on Build GRCh38
rs530639365 Chr.11: 104995764-104995764 on Build GRCh38
rs771432226 Chr.11: 104995839-104995839 on Build GRCh38
rs772675370 Chr.11: 104995860-104995860 on Build GRCh38
rs369985181 Chr.11: 104995779-104995779 on Build GRCh38
rs780000213 Chr.11: 104995726-104995726 on Build GRCh38
rs996397530 Chr.11: 104995825-104995825 on Build GRCh38
rs977378511 Chr.11: 104995704-104995704 on Build GRCh38
rs976337392 Chr.11: 104995736-104995736 on Build GRCh38
rs377647933 Chr.11: 104995856-104995856 on Build GRCh38
rs150806636 Chr.11: 104995793-104995793 on Build GRCh38
rs540819 Chr.11: 104995734-104995734 on Build GRCh38
rs146387103 Chr.11: 104995756-104995756 on Build GRCh38
rs750199864 Chr.11: 104995722-104995722 on Build GRCh38
rs761406463 Chr.11: 104995875-104995875 on Build GRCh38
rs375833242 Chr.11: 104995747-104995747 on Build GRCh38
rs773500942 Chr.11: 104995750-104995750 on Build GRCh38
rs754909891 Chr.11: 104995818-104995818 on Build GRCh38
rs138021690 Chr.11: 104995802-104995802 on Build GRCh38
rs768738083 Chr.11: 104995733-104995733 on Build GRCh38
rs751555513 Chr.11: 104995813-104995813 on Build GRCh38
rs781292126 Chr.11: 104995824-104995824 on Build GRCh38
rs377165670 Chr.11: 104995730-104995730 on Build GRCh38
rs371132766 Chr.11: 104995873-104995873 on Build GRCh38

View Full Product Details