Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119715335-119715608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAATS1
Entrez Gene ID
89876
Gene Name
MYCBP associated and testis expressed 1
Gene Aliases
AAT1, AAT1alpha, C3orf15, CaM-IP2, SPATA26
UniGene
Hs.341906
Gene Chromosome Location
Chr.3: 119703022-119767102 on Build GRCh38
Transcripts
NM_001320316.1 NM_001320317.1 NM_001320318.1 NM_033364.3 XM_011513277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3914637 c.545C>T p.S182F Chr.3: 119715606-119715606 on Build GRCh38
1036777 c.534T>C p.T178T Chr.3: 119715595-119715595 on Build GRCh38
4112567 c.514A>G p.T172A Chr.3: 119715575-119715575 on Build GRCh38
6163073 c.530C>T p.S177F Chr.3: 119715591-119715591 on Build GRCh38
3914638 c.545C>T p.S182F Chr.3: 119715606-119715606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150795242 Chr.3: 119715579-119715579 on Build GRCh38
rs998468734 Chr.3: 119715460-119715460 on Build GRCh38
rs756723844 Chr.3: 119715514-119715514 on Build GRCh38
rs778677534 Chr.3: 119715535-119715535 on Build GRCh38
rs1031262951 Chr.3: 119715494-119715494 on Build GRCh38
rs375655115 Chr.3: 119715522-119715522 on Build GRCh38
rs374047605 Chr.3: 119715553-119715553 on Build GRCh38
rs367898250 Chr.3: 119715554-119715554 on Build GRCh38
rs528858712 Chr.3: 119715458-119715458 on Build GRCh38
rs763065173 Chr.3: 119715393-119715393 on Build GRCh38
rs753673815 Chr.3: 119715392-119715392 on Build GRCh38
rs370858430 Chr.3: 119715526-119715526 on Build GRCh38
rs778563874 Chr.3: 119715534-119715534 on Build GRCh38
rs764330129 Chr.3: 119715395-119715395 on Build GRCh38
rs951168502 Chr.3: 119715573-119715573 on Build GRCh38
rs780540554 Chr.3: 119715515-119715515 on Build GRCh38
rs751791444 Chr.3: 119715576-119715576 on Build GRCh38
rs201703483 Chr.3: 119715559-119715559 on Build GRCh38
rs543494111 Chr.3: 119715369-119715369 on Build GRCh38
rs750960944 Chr.3: 119715412-119715412 on Build GRCh38
rs771601553 Chr.3: 119715546-119715546 on Build GRCh38
rs999606253 Chr.3: 119715480-119715480 on Build GRCh38
rs576249462 Chr.3: 119715359-119715359 on Build GRCh38
rs757223529 Chr.3: 119715479-119715479 on Build GRCh38
rs766158734 Chr.3: 119715376-119715376 on Build GRCh38
rs747771788 Chr.3: 119715540-119715540 on Build GRCh38
rs373269234 Chr.3: 119715565-119715565 on Build GRCh38
rs142224777 Chr.3: 119715564-119715564 on Build GRCh38
rs180738357 Chr.3: 119715403-119715403 on Build GRCh38
rs1029700382 Chr.3: 119715492-119715492 on Build GRCh38
rs781187516 Chr.3: 119715405-119715405 on Build GRCh38
rs770905232 Chr.3: 119715555-119715555 on Build GRCh38
rs1041253378 Chr.3: 119715366-119715366 on Build GRCh38
rs751732861 Chr.3: 119715402-119715402 on Build GRCh38
rs955343535 Chr.3: 119715533-119715533 on Build GRCh38
rs375483032 Chr.3: 119715556-119715556 on Build GRCh38
rs1027064201 Chr.3: 119715580-119715580 on Build GRCh38

View Full Product Details