Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50739771-50740044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACR
Entrez Gene ID
49
Gene Name
acrosin
Gene Aliases
-
UniGene
Hs.370870
Gene Chromosome Location
Chr.22: 50738224-50745299 on Build GRCh38
Transcripts
NM_001097.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
479143 c.554T>C p.I185T Chr.22: 50739966-50739966 on Build GRCh38
4399339 c.419G>A p.G140E Chr.22: 50739831-50739831 on Build GRCh38
4905224 c.458C>T p.S153L Chr.22: 50739870-50739870 on Build GRCh38
4438298 c.394G>C p.E132Q Chr.22: 50739806-50739806 on Build GRCh38
3555866 c.426C>T p.D142D Chr.22: 50739838-50739838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754127030 Chr.22: 50739950-50739950 on Build GRCh38
rs368795913 Chr.22: 50739966-50739966 on Build GRCh38
rs140364806 Chr.22: 50739858-50739858 on Build GRCh38
rs764169031 Chr.22: 50739952-50739952 on Build GRCh38
rs201620831 Chr.22: 50739852-50739852 on Build GRCh38
rs750510857 Chr.22: 50739958-50739958 on Build GRCh38
rs547219491 Chr.22: 50739987-50739987 on Build GRCh38
rs766337786 Chr.22: 50740024-50740024 on Build GRCh38
rs768599127 Chr.22: 50739871-50739871 on Build GRCh38
rs781212788 Chr.22: 50739977-50739977 on Build GRCh38
rs761053255 Chr.22: 50740003-50740003 on Build GRCh38
rs773984711 Chr.22: 50739891-50739891 on Build GRCh38
rs765965924 Chr.22: 50739819-50739819 on Build GRCh38
rs547752864 Chr.22: 50740008-50740008 on Build GRCh38
rs768141356 Chr.22: 50739907-50739907 on Build GRCh38
rs764583729 Chr.22: 50740005-50740005 on Build GRCh38
rs529658418 Chr.22: 50739999-50739999 on Build GRCh38
rs759056819 Chr.22: 50739892-50739892 on Build GRCh38
rs774080258 Chr.22: 50739992-50739992 on Build GRCh38
rs376947740 Chr.22: 50739953-50739953 on Build GRCh38
rs368710046 Chr.22: 50739870-50739870 on Build GRCh38
rs768051648 Chr.22: 50739829-50739829 on Build GRCh38
rs543728233 Chr.22: 50739802-50739802 on Build GRCh38
rs867063389 Chr.22: 50739929-50739929 on Build GRCh38
rs745401505 Chr.22: 50739997-50739997 on Build GRCh38
rs755978747 Chr.22: 50739985-50739985 on Build GRCh38
rs748013634 Chr.22: 50739982-50739982 on Build GRCh38
rs202122603 Chr.22: 50739885-50739885 on Build GRCh38
rs748010379 Chr.22: 50739878-50739878 on Build GRCh38
rs765479356 Chr.22: 50740011-50740011 on Build GRCh38
rs753117359 Chr.22: 50739836-50739836 on Build GRCh38
rs1053770577 Chr.22: 50739989-50739989 on Build GRCh38
rs760115224 Chr.22: 50739828-50739828 on Build GRCh38
rs776109873 Chr.22: 50740001-50740001 on Build GRCh38
rs750598701 Chr.22: 50740012-50740012 on Build GRCh38
rs752153585 Chr.22: 50739822-50739822 on Build GRCh38
rs371591031 Chr.22: 50739856-50739856 on Build GRCh38
rs763011946 Chr.22: 50739914-50739914 on Build GRCh38
rs762663990 Chr.22: 50739889-50739889 on Build GRCh38
rs756365879 Chr.22: 50739847-50739847 on Build GRCh38
rs764339107 Chr.22: 50739942-50739942 on Build GRCh38
rs113204118 Chr.22: 50739986-50739986 on Build GRCh38
rs182889325 Chr.22: 50739991-50739991 on Build GRCh38
rs1064735 Chr.22: 50739910-50739910 on Build GRCh38
rs753102410 Chr.22: 50739918-50739918 on Build GRCh38
rs758522868 Chr.22: 50740023-50740023 on Build GRCh38
rs753955522 Chr.22: 50739848-50739848 on Build GRCh38
rs769428134 Chr.22: 50739879-50739879 on Build GRCh38
rs776876305 Chr.22: 50740007-50740007 on Build GRCh38

View Full Product Details