Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123084479-123084750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
UniGene
Hs.591949
Gene Chromosome Location
Chr.11: 123072006-123195305 on Build GRCh38
Transcripts
NM_024769.3 XM_011542998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704730 c.244C>T p.R82* Chr.11: 123084656-123084656 on Build GRCh38
3444499 c.232G>A p.E78K Chr.11: 123084668-123084668 on Build GRCh38
1676531 c.287T>A p.L96* Chr.11: 123084613-123084613 on Build GRCh38
4189335 c.250G>A p.A84T Chr.11: 123084650-123084650 on Build GRCh38
6223303 c.205C>T p.R69C Chr.11: 123084695-123084695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587776964 Chr.11: 123084670-123084670 on Build GRCh38
rs781327868 Chr.11: 123084538-123084538 on Build GRCh38
rs749127591 Chr.11: 123084659-123084659 on Build GRCh38
rs367894332 Chr.11: 123084648-123084648 on Build GRCh38
rs370904077 Chr.11: 123084576-123084576 on Build GRCh38
rs750651117 Chr.11: 123084641-123084641 on Build GRCh38
rs377683000 Chr.11: 123084503-123084503 on Build GRCh38
rs866004039 Chr.11: 123084625-123084625 on Build GRCh38
rs183962570 Chr.11: 123084632-123084632 on Build GRCh38
rs35602197 Chr.11: 123084562-123084563 on Build GRCh38
rs751630847 Chr.11: 123084655-123084655 on Build GRCh38
rs867962634 Chr.11: 123084671-123084671 on Build GRCh38
rs776048110 Chr.11: 123084597-123084597 on Build GRCh38
rs368880095 Chr.11: 123084665-123084665 on Build GRCh38
rs138315099 Chr.11: 123084627-123084627 on Build GRCh38
rs2276348 Chr.11: 123084694-123084694 on Build GRCh38
rs769237709 Chr.11: 123084701-123084701 on Build GRCh38
rs141499494 Chr.11: 123084542-123084542 on Build GRCh38
rs745428396 Chr.11: 123084592-123084592 on Build GRCh38
rs1043218913 Chr.11: 123084660-123084660 on Build GRCh38
rs758504860 Chr.11: 123084535-123084535 on Build GRCh38
rs748967980 Chr.11: 123084547-123084547 on Build GRCh38
rs756079825 Chr.11: 123084656-123084656 on Build GRCh38
rs765488751 Chr.11: 123084728-123084728 on Build GRCh38
rs371011814 Chr.11: 123084693-123084693 on Build GRCh38
rs771632957 Chr.11: 123084683-123084683 on Build GRCh38
rs150466973 Chr.11: 123084602-123084602 on Build GRCh38
rs777061354 Chr.11: 123084613-123084613 on Build GRCh38
rs748093808 Chr.11: 123084540-123084540 on Build GRCh38
rs750549964 Chr.11: 123084531-123084531 on Build GRCh38
rs773488512 Chr.11: 123084610-123084610 on Build GRCh38
rs764453599 Chr.11: 123084612-123084612 on Build GRCh38
rs758618133 Chr.11: 123084649-123084649 on Build GRCh38
rs762287014 Chr.11: 123084721-123084721 on Build GRCh38
rs776956196 Chr.11: 123084710-123084710 on Build GRCh38
rs771535184 Chr.11: 123084593-123084593 on Build GRCh38
rs746453314 Chr.11: 123084695-123084695 on Build GRCh38
rs770645005 Chr.11: 123084548-123084548 on Build GRCh38
rs773823695 Chr.11: 123084578-123084578 on Build GRCh38
rs1001764516 Chr.11: 123084727-123084727 on Build GRCh38
rs879186276 Chr.11: 123084555-123084555 on Build GRCh38
rs148377321 Chr.11: 123084517-123084517 on Build GRCh38
rs886886089 Chr.11: 123084607-123084607 on Build GRCh38
rs374703230 Chr.11: 123084657-123084657 on Build GRCh38
rs546517283 Chr.11: 123084543-123084543 on Build GRCh38
rs587776967 Chr.11: 123084529-123084529 on Build GRCh38
rs778937363 Chr.11: 123084505-123084505 on Build GRCh38
rs745519749 Chr.11: 123084675-123084675 on Build GRCh38

View Full Product Details