Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223393312-223393557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC185
Entrez Gene ID
164127
Gene Name
coiled-coil domain containing 185
Gene Aliases
C1orf65
UniGene
Hs.192090
Gene Chromosome Location
Chr.1: 223393373-223395470 on Build GRCh38
Transcripts
NM_152610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1320452 c.59G>A p.R20Q Chr.1: 223393534-223393534 on Build GRCh38
1601695 c.37C>A p.R13R Chr.1: 223393512-223393512 on Build GRCh38
3750901 c.42T>C p.D14D Chr.1: 223393517-223393517 on Build GRCh38
4667422 c.23delC p.Q9fs*77 Chr.1: 223393498-223393498 on Build GRCh38
3689370 c.56C>T p.P19L Chr.1: 223393531-223393531 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045885352 Chr.1: 223393350-223393350 on Build GRCh38
rs753470706 Chr.1: 223393533-223393533 on Build GRCh38
rs377273038 Chr.1: 223393374-223393374 on Build GRCh38
rs555558273 Chr.1: 223393395-223393395 on Build GRCh38
rs750227441 Chr.1: 223393516-223393516 on Build GRCh38
rs372910030 Chr.1: 223393445-223393445 on Build GRCh38
rs75945379 Chr.1: 223393534-223393534 on Build GRCh38
rs905578584 Chr.1: 223393358-223393358 on Build GRCh38
rs769629901 Chr.1: 223393518-223393530 on Build GRCh38
rs1025789084 Chr.1: 223393498-223393498 on Build GRCh38
rs111306873 Chr.1: 223393517-223393517 on Build GRCh38
rs763086399 Chr.1: 223393477-223393477 on Build GRCh38
rs917716912 Chr.1: 223393347-223393347 on Build GRCh38
rs866707986 Chr.1: 223393491-223393491 on Build GRCh38
rs1027302343 Chr.1: 223393375-223393375 on Build GRCh38
rs118102924 Chr.1: 223393380-223393380 on Build GRCh38
rs770154745 Chr.1: 223393457-223393457 on Build GRCh38
rs755957555 Chr.1: 223393527-223393527 on Build GRCh38
rs528074660 Chr.1: 223393474-223393474 on Build GRCh38
rs994663365 Chr.1: 223393471-223393471 on Build GRCh38
rs1001205476 Chr.1: 223393372-223393372 on Build GRCh38
rs375208182 Chr.1: 223393400-223393400 on Build GRCh38
rs959062031 Chr.1: 223393334-223393334 on Build GRCh38
rs767232747 Chr.1: 223393514-223393514 on Build GRCh38
rs57755601 Chr.1: 223393452-223393452 on Build GRCh38
rs922680610 Chr.1: 223393406-223393406 on Build GRCh38
rs534062463 Chr.1: 223393394-223393394 on Build GRCh38
rs202074898 Chr.1: 223393458-223393458 on Build GRCh38
rs1005747677 Chr.1: 223393512-223393512 on Build GRCh38
rs372220702 Chr.1: 223393538-223393538 on Build GRCh38
rs913097305 Chr.1: 223393496-223393496 on Build GRCh38
rs569169878 Chr.1: 223393348-223393348 on Build GRCh38
rs976912900 Chr.1: 223393402-223393402 on Build GRCh38
rs990514560 Chr.1: 223393346-223393346 on Build GRCh38
rs374264495 Chr.1: 223393513-223393513 on Build GRCh38
rs576486474 Chr.1: 223393419-223393419 on Build GRCh38
rs563608413 Chr.1: 223393426-223393426 on Build GRCh38
rs766009267 Chr.1: 223393531-223393531 on Build GRCh38
rs1053440884 Chr.1: 223393475-223393475 on Build GRCh38
rs539454389 Chr.1: 223393365-223393365 on Build GRCh38
rs774294698 Chr.1: 223393509-223393509 on Build GRCh38
rs764126570 Chr.1: 223393504-223393504 on Build GRCh38

View Full Product Details