Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54724937-54725182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML6
Entrez Gene ID
400954
Gene Name
echinoderm microtubule associated protein like 6
Gene Aliases
-
UniGene
Hs.656692
Gene Chromosome Location
Chr.2: 54723467-54972908 on Build GRCh38
Transcripts
NM_001039753.2 XM_017004098.1 XM_017004099.1 XM_017004100.1 XM_017004102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4171179 c.7G>T p.D3Y Chr.2: 54725068-54725068 on Build GRCh38
4649997 c.18G>T p.A6A Chr.2: 54725079-54725079 on Build GRCh38
5656493 c.86A>G p.Y29C Chr.2: 54725147-54725147 on Build GRCh38
3714211 c.45G>T p.W15C Chr.2: 54725106-54725106 on Build GRCh38
3396484 c.50A>T p.Y17F Chr.2: 54725111-54725111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752098287 Chr.2: 54725155-54725155 on Build GRCh38
rs541484635 Chr.2: 54725003-54725003 on Build GRCh38
rs554556276 Chr.2: 54725016-54725016 on Build GRCh38
rs752894375 Chr.2: 54725095-54725095 on Build GRCh38
rs771598167 Chr.2: 54725022-54725022 on Build GRCh38
rs930284059 Chr.2: 54725024-54725024 on Build GRCh38
rs962648125 Chr.2: 54724982-54724982 on Build GRCh38
rs369146275 Chr.2: 54725076-54725076 on Build GRCh38
rs969322792 Chr.2: 54725007-54725007 on Build GRCh38
rs769738994 Chr.2: 54725130-54725130 on Build GRCh38
rs950669183 Chr.2: 54725154-54725154 on Build GRCh38
rs900254169 Chr.2: 54725053-54725053 on Build GRCh38
rs1030213338 Chr.2: 54724963-54724963 on Build GRCh38
rs759267390 Chr.2: 54725047-54725047 on Build GRCh38
rs112552344 Chr.2: 54725027-54725027 on Build GRCh38
rs576214877 Chr.2: 54725004-54725004 on Build GRCh38
rs539190629 Chr.2: 54725035-54725035 on Build GRCh38
rs995808645 Chr.2: 54725066-54725066 on Build GRCh38
rs868037094 Chr.2: 54725059-54725059 on Build GRCh38
rs143216688 Chr.2: 54724978-54724978 on Build GRCh38
rs758523258 Chr.2: 54725133-54725133 on Build GRCh38
rs559043648 Chr.2: 54724980-54724980 on Build GRCh38
rs562435762 Chr.2: 54725104-54725104 on Build GRCh38
rs773100562 Chr.2: 54725031-54725031 on Build GRCh38
rs999478234 Chr.2: 54725125-54725125 on Build GRCh38
rs531572203 Chr.2: 54725141-54725141 on Build GRCh38
rs572423410 Chr.2: 54724992-54724992 on Build GRCh38
rs1030858692 Chr.2: 54724980-54724981 on Build GRCh38
rs1027666696 Chr.2: 54725120-54725120 on Build GRCh38
rs1051461612 Chr.2: 54725097-54725097 on Build GRCh38
rs766160944 Chr.2: 54725036-54725036 on Build GRCh38
rs954658025 Chr.2: 54724966-54724966 on Build GRCh38
rs574449759 Chr.2: 54725030-54725030 on Build GRCh38
rs753606825 Chr.2: 54725043-54725043 on Build GRCh38
rs765451011 Chr.2: 54725085-54725085 on Build GRCh38
rs890351019 Chr.2: 54725107-54725107 on Build GRCh38

View Full Product Details