Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135115517-135115742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
UniGene
Hs.648616
Gene Chromosome Location
Chr.7: 134985169-135135782 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
UniGene
Hs.643113
Gene Chromosome Location
Chr.7: 135092303-135170826 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792053 c.2342+7C>G p.? Chr.7: 135115618-135115618 on Build GRCh38
452393 c.2269A>G p.S757G Chr.7: 135115538-135115538 on Build GRCh38
7426629 c.2494C>T p.Q832* Chr.7: 135115598-135115598 on Build GRCh38
7426628 c.2329C>T p.Q777* Chr.7: 135115598-135115598 on Build GRCh38
5792054 c.2507+7C>G p.? Chr.7: 135115618-135115618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114250710 Chr.7: 135115651-135115651 on Build GRCh38
rs371770007 Chr.7: 135115638-135115641 on Build GRCh38
rs576710817 Chr.7: 135115628-135115628 on Build GRCh38
rs572746285 Chr.7: 135115693-135115693 on Build GRCh38
rs771017532 Chr.7: 135115654-135115655 on Build GRCh38
rs868252390 Chr.7: 135115641-135115641 on Build GRCh38
rs893275863 Chr.7: 135115685-135115685 on Build GRCh38
rs754445096 Chr.7: 135115631-135115631 on Build GRCh38
rs1049908497 Chr.7: 135115721-135115721 on Build GRCh38
rs779054463 Chr.7: 135115568-135115569 on Build GRCh38
rs963686153 Chr.7: 135115620-135115620 on Build GRCh38
rs932840286 Chr.7: 135115717-135115717 on Build GRCh38
rs746442579 Chr.7: 135115684-135115684 on Build GRCh38
rs1046469020 Chr.7: 135115694-135115695 on Build GRCh38
rs755073596 Chr.7: 135115646-135115646 on Build GRCh38
rs565589064 Chr.7: 135115661-135115661 on Build GRCh38
rs556838766 Chr.7: 135115576-135115576 on Build GRCh38
rs978151145 Chr.7: 135115552-135115552 on Build GRCh38
rs750731396 Chr.7: 135115537-135115537 on Build GRCh38

View Full Product Details