Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13388994-13389219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
UniGene
Hs.332382
Gene Chromosome Location
Chr.11: 13387996-13463297 on Build GRCh38
Transcripts
NM_001297741.1 NM_001297742.1 NM_032320.6 XM_017018405.1 XM_017018406.1 XM_017018407.1 XM_017018408.1 XM_017018409.1 XM_017018410.1 XM_017018411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
458686 c.1205G>A p.R402Q Chr.11: 13389054-13389054 on Build GRCh38
1627876 c.1118-6_1118-3delTTTC p.? Chr.11: 13389144-13389147 on Build GRCh38
6224028 c.1162C>T p.R388C Chr.11: 13389097-13389097 on Build GRCh38
6613296 c.1156G>A p.G386R Chr.11: 13389103-13389103 on Build GRCh38
7104778 c.1163G>T p.R388L Chr.11: 13389096-13389096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147243996 Chr.11: 13389118-13389118 on Build GRCh38
rs3833472 Chr.11: 13389144-13389147 on Build GRCh38
rs755916930 Chr.11: 13389057-13389057 on Build GRCh38
rs948304085 Chr.11: 13389070-13389070 on Build GRCh38
rs764047272 Chr.11: 13389062-13389062 on Build GRCh38
rs766656420 Chr.11: 13389045-13389045 on Build GRCh38
rs750003509 Chr.11: 13389116-13389116 on Build GRCh38
rs753752344 Chr.11: 13389096-13389096 on Build GRCh38
rs759997243 Chr.11: 13389151-13389152 on Build GRCh38
rs753180265 Chr.11: 13389153-13389153 on Build GRCh38
rs375824034 Chr.11: 13389154-13389157 on Build GRCh38
rs563825040 Chr.11: 13389102-13389102 on Build GRCh38
rs778015480 Chr.11: 13389157-13389157 on Build GRCh38
rs765763753 Chr.11: 13389162-13389162 on Build GRCh38
rs781693674 Chr.11: 13389177-13389177 on Build GRCh38
rs144141685 Chr.11: 13389032-13389032 on Build GRCh38
rs748571680 Chr.11: 13389188-13389188 on Build GRCh38
rs56274863 Chr.11: 13389103-13389103 on Build GRCh38
rs753011483 Chr.11: 13389175-13389176 on Build GRCh38
rs956138191 Chr.11: 13389173-13389173 on Build GRCh38
rs990107334 Chr.11: 13389176-13389176 on Build GRCh38
rs75019394 Chr.11: 13389054-13389054 on Build GRCh38
rs769163052 Chr.11: 13389164-13389164 on Build GRCh38
rs1007917541 Chr.11: 13389155-13389155 on Build GRCh38
rs773380514 Chr.11: 13389023-13389023 on Build GRCh38
rs752423002 Chr.11: 13389055-13389055 on Build GRCh38
rs758725248 Chr.11: 13389154-13389154 on Build GRCh38

View Full Product Details