Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62253002-62253273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK4
Entrez Gene ID
163782
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
ANKRD38, dJ1078M7.1
UniGene
Hs.283398
Gene Chromosome Location
Chr.1: 62236165-62319414 on Build GRCh38
Transcripts
NM_001320269.1 NM_181712.4 XM_011540843.2 XM_011540845.2 XM_017000483.1 XM_017000484.1 XM_017000485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5491130 c.2541C>T p.G847G Chr.1: 62253208-62253208 on Build GRCh38
4393231 c.2541C>A p.G847G Chr.1: 62253208-62253208 on Build GRCh38
6690032 c.2656G>T p.G886* Chr.1: 62253093-62253093 on Build GRCh38
3865953 c.2553G>A p.V851V Chr.1: 62253196-62253196 on Build GRCh38
681559 c.2632G>A p.V878I Chr.1: 62253117-62253117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771067968 Chr.1: 62253202-62253202 on Build GRCh38
rs779807184 Chr.1: 62253080-62253080 on Build GRCh38
rs1057386990 Chr.1: 62253089-62253089 on Build GRCh38
rs150211365 Chr.1: 62253141-62253141 on Build GRCh38
rs755414930 Chr.1: 62253098-62253098 on Build GRCh38
rs747022382 Chr.1: 62253200-62253200 on Build GRCh38
rs766941812 Chr.1: 62253053-62253053 on Build GRCh38
rs868053826 Chr.1: 62253219-62253219 on Build GRCh38
rs781337033 Chr.1: 62253206-62253206 on Build GRCh38
rs746099288 Chr.1: 62253207-62253207 on Build GRCh38
rs369699713 Chr.1: 62253170-62253170 on Build GRCh38
rs777093316 Chr.1: 62253209-62253210 on Build GRCh38
rs755712183 Chr.1: 62253176-62253176 on Build GRCh38
rs377521746 Chr.1: 62253165-62253165 on Build GRCh38
rs745763282 Chr.1: 62253166-62253166 on Build GRCh38
rs751522890 Chr.1: 62253049-62253049 on Build GRCh38
rs765990772 Chr.1: 62253227-62253227 on Build GRCh38
rs771024675 Chr.1: 62253124-62253124 on Build GRCh38
rs753557460 Chr.1: 62253161-62253161 on Build GRCh38
rs774327899 Chr.1: 62253052-62253052 on Build GRCh38
rs368721566 Chr.1: 62253024-62253024 on Build GRCh38
rs922200040 Chr.1: 62253221-62253221 on Build GRCh38
rs745635707 Chr.1: 62253102-62253102 on Build GRCh38
rs570900175 Chr.1: 62253096-62253096 on Build GRCh38
rs773222515 Chr.1: 62253216-62253216 on Build GRCh38
rs775122850 Chr.1: 62253036-62253036 on Build GRCh38
rs149058627 Chr.1: 62253192-62253192 on Build GRCh38
rs760044201 Chr.1: 62253225-62253225 on Build GRCh38
rs148060294 Chr.1: 62253208-62253208 on Build GRCh38
rs755326990 Chr.1: 62253162-62253162 on Build GRCh38
rs776871936 Chr.1: 62253130-62253130 on Build GRCh38
rs147615893 Chr.1: 62253047-62253047 on Build GRCh38
rs762778995 Chr.1: 62253038-62253038 on Build GRCh38
rs747158846 Chr.1: 62253117-62253117 on Build GRCh38
rs760261691 Chr.1: 62253153-62253153 on Build GRCh38
rs766678111 Chr.1: 62253031-62253031 on Build GRCh38
rs113961902 Chr.1: 62253151-62253151 on Build GRCh38
rs749881567 Chr.1: 62253149-62253149 on Build GRCh38
rs753465407 Chr.1: 62253230-62253230 on Build GRCh38
rs772698544 Chr.1: 62253116-62253116 on Build GRCh38
rs143679738 Chr.1: 62253155-62253155 on Build GRCh38
rs142552073 Chr.1: 62253131-62253131 on Build GRCh38
rs774212372 Chr.1: 62253136-62253136 on Build GRCh38
rs200229473 Chr.1: 62253132-62253132 on Build GRCh38
rs982437548 Chr.1: 62253237-62253237 on Build GRCh38
rs750017708 Chr.1: 62253063-62253063 on Build GRCh38
rs368901402 Chr.1: 62253064-62253064 on Build GRCh38
rs540592145 Chr.1: 62253176-62253176 on Build GRCh38
rs759247190 Chr.1: 62253248-62253248 on Build GRCh38
rs199721081 Chr.1: 62253142-62253142 on Build GRCh38
rs145530313 Chr.1: 62253099-62253099 on Build GRCh38
rs771874857 Chr.1: 62253212-62253212 on Build GRCh38

View Full Product Details