Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54842916-54843185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCH1
Entrez Gene ID
2643
Gene Name
GTP cyclohydrolase 1
Gene Aliases
DYT14, DYT5, DYT5a, GCH, GTP-CH-1, GTPCH1, HPABH4B
UniGene
Hs.86724
Gene Chromosome Location
Chr.14: 54842005-54902824 on Build GRCh38
Transcripts
NM_000161.2 NM_001024024.1 NM_001024070.1 NM_001024071.1 XM_017021218.1

SNP Detail

SNP ID Chromosome Location
rs770558029 Chr.14: 54843002-54843002 on Build GRCh38
rs939635665 Chr.14: 54843087-54843087 on Build GRCh38
rs941682193 Chr.14: 54843140-54843141 on Build GRCh38
rs868303976 Chr.14: 54843044-54843044 on Build GRCh38
rs773107365 Chr.14: 54843076-54843076 on Build GRCh38
rs189352198 Chr.14: 54843028-54843028 on Build GRCh38
rs549379993 Chr.14: 54842975-54842975 on Build GRCh38
rs144676716 Chr.14: 54842960-54842961 on Build GRCh38
rs376668776 Chr.14: 54843031-54843031 on Build GRCh38
rs758614226 Chr.14: 54843022-54843022 on Build GRCh38
rs867220379 Chr.14: 54842984-54842984 on Build GRCh38
rs772782531 Chr.14: 54842999-54842999 on Build GRCh38
rs879179089 Chr.14: 54843134-54843134 on Build GRCh38
rs930893834 Chr.14: 54843116-54843116 on Build GRCh38
rs568895596 Chr.14: 54843018-54843018 on Build GRCh38
rs770712450 Chr.14: 54843034-54843034 on Build GRCh38
rs143961472 Chr.14: 54842973-54842973 on Build GRCh38
rs1055904097 Chr.14: 54842960-54842960 on Build GRCh38
rs748793695 Chr.14: 54843051-54843051 on Build GRCh38
rs995386735 Chr.14: 54842959-54842959 on Build GRCh38
rs908913666 Chr.14: 54843124-54843125 on Build GRCh38
rs777628224 Chr.14: 54843036-54843036 on Build GRCh38
rs930222058 Chr.14: 54843117-54843117 on Build GRCh38
rs183455779 Chr.14: 54843003-54843003 on Build GRCh38
rs1023705665 Chr.14: 54843065-54843065 on Build GRCh38
rs769144102 Chr.14: 54842998-54842998 on Build GRCh38
rs768865732 Chr.14: 54843083-54843083 on Build GRCh38
rs145762799 Chr.14: 54843123-54843125 on Build GRCh38
rs983094016 Chr.14: 54843123-54843128 on Build GRCh38
rs955141614 Chr.14: 54843042-54843042 on Build GRCh38
rs140865143 Chr.14: 54843067-54843067 on Build GRCh38
rs150937402 Chr.14: 54843039-54843039 on Build GRCh38
rs764569623 Chr.14: 54843007-54843007 on Build GRCh38
rs776957978 Chr.14: 54843015-54843015 on Build GRCh38
rs1014282544 Chr.14: 54842983-54842983 on Build GRCh38
rs773983177 Chr.14: 54843042-54843042 on Build GRCh38
rs918838734 Chr.14: 54843116-54843117 on Build GRCh38
rs1036983132 Chr.14: 54842947-54842947 on Build GRCh38
rs899341447 Chr.14: 54842957-54842957 on Build GRCh38
rs920823865 Chr.14: 54843109-54843109 on Build GRCh38
rs180979582 Chr.14: 54843001-54843001 on Build GRCh38
rs777063292 Chr.14: 54843086-54843086 on Build GRCh38
rs868459642 Chr.14: 54842946-54842946 on Build GRCh38
rs370056236 Chr.14: 54843021-54843021 on Build GRCh38
rs1038747682 Chr.14: 54842955-54842955 on Build GRCh38
rs765167079 Chr.14: 54843017-54843017 on Build GRCh38
rs762415774 Chr.14: 54843016-54843016 on Build GRCh38
rs971340420 Chr.14: 54843079-54843079 on Build GRCh38
rs748137436 Chr.14: 54842989-54842989 on Build GRCh38
rs556250070 Chr.14: 54843032-54843032 on Build GRCh38
rs769757099 Chr.14: 54843066-54843066 on Build GRCh38
rs554432105 Chr.14: 54843094-54843094 on Build GRCh38
rs756091241 Chr.14: 54843035-54843035 on Build GRCh38
rs760998949 Chr.14: 54843004-54843004 on Build GRCh38

View Full Product Details