Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72221015-72221272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLR2
Entrez Gene ID
2350
Gene Name
folate receptor beta
Gene Aliases
BETA-HFR, FBP, FBP/PL-1, FR-BETA, FR-P3
UniGene
Hs.433159
Gene Chromosome Location
Chr.11: 72216775-72221950 on Build GRCh38
Transcripts
NM_000803.4 NM_001113534.1 NM_001113535.1 NM_001113536.1 XM_005273856.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3710422 c.421C>A p.H141N Chr.11: 72221257-72221257 on Build GRCh38
3452985 c.334C>T p.Q112* Chr.11: 72221053-72221053 on Build GRCh38
1207386 c.356G>A p.R119H Chr.11: 72221192-72221192 on Build GRCh38
2016102 c.367T>G p.F123V Chr.11: 72221203-72221203 on Build GRCh38
4491539 c.381C>T p.P127P Chr.11: 72221217-72221217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377722879 Chr.11: 72221156-72221156 on Build GRCh38
rs202185342 Chr.11: 72221085-72221085 on Build GRCh38
rs752497032 Chr.11: 72221042-72221042 on Build GRCh38
rs760392076 Chr.11: 72221086-72221094 on Build GRCh38
rs779883926 Chr.11: 72221254-72221254 on Build GRCh38
rs571762536 Chr.11: 72221080-72221080 on Build GRCh38
rs769483853 Chr.11: 72221086-72221086 on Build GRCh38
rs771588531 Chr.11: 72221076-72221077 on Build GRCh38
rs1042498602 Chr.11: 72221105-72221105 on Build GRCh38
rs759805548 Chr.11: 72221220-72221220 on Build GRCh38
rs1803569 Chr.11: 72221255-72221255 on Build GRCh38
rs141425716 Chr.11: 72221240-72221240 on Build GRCh38
rs765342233 Chr.11: 72221145-72221145 on Build GRCh38
rs568731068 Chr.11: 72221133-72221133 on Build GRCh38
rs770391415 Chr.11: 72221069-72221069 on Build GRCh38
rs748662206 Chr.11: 72221066-72221067 on Build GRCh38
rs549721589 Chr.11: 72221075-72221075 on Build GRCh38
rs141294007 Chr.11: 72221193-72221193 on Build GRCh38
rs199974430 Chr.11: 72221078-72221078 on Build GRCh38
rs369297388 Chr.11: 72221157-72221157 on Build GRCh38
rs779399069 Chr.11: 72221064-72221064 on Build GRCh38
rs193165102 Chr.11: 72221192-72221192 on Build GRCh38
rs367823426 Chr.11: 72221069-72221069 on Build GRCh38
rs1000842218 Chr.11: 72221125-72221125 on Build GRCh38
rs763868667 Chr.11: 72221097-72221098 on Build GRCh38
rs759152691 Chr.11: 72221072-72221072 on Build GRCh38
rs753183270 Chr.11: 72221152-72221152 on Build GRCh38
rs138451850 Chr.11: 72221183-72221183 on Build GRCh38
rs148292921 Chr.11: 72221236-72221236 on Build GRCh38
rs771923158 Chr.11: 72221205-72221205 on Build GRCh38
rs749773371 Chr.11: 72221063-72221063 on Build GRCh38
rs768312002 Chr.11: 72221088-72221088 on Build GRCh38
rs759609391 Chr.11: 72221134-72221134 on Build GRCh38
rs774146276 Chr.11: 72221091-72221091 on Build GRCh38
rs374592973 Chr.11: 72221101-72221101 on Build GRCh38
rs780220815 Chr.11: 72221061-72221061 on Build GRCh38
rs761543780 Chr.11: 72221095-72221095 on Build GRCh38
rs750713005 Chr.11: 72221053-72221053 on Build GRCh38
rs773715246 Chr.11: 72221069-72221070 on Build GRCh38
rs371973215 Chr.11: 72221158-72221158 on Build GRCh38
rs371396761 Chr.11: 72221191-72221191 on Build GRCh38
rs143308196 Chr.11: 72221233-72221233 on Build GRCh38
rs556999096 Chr.11: 72221087-72221087 on Build GRCh38
rs768031158 Chr.11: 72221250-72221250 on Build GRCh38
rs1051046742 Chr.11: 72221139-72221139 on Build GRCh38
rs757571036 Chr.11: 72221162-72221162 on Build GRCh38
rs767575485 Chr.11: 72221048-72221048 on Build GRCh38
rs752060276 Chr.11: 72221239-72221239 on Build GRCh38
rs375901970 Chr.11: 72221040-72221040 on Build GRCh38
rs1039785844 Chr.11: 72221109-72221109 on Build GRCh38
rs781644219 Chr.11: 72221082-72221082 on Build GRCh38
rs998532093 Chr.11: 72221120-72221120 on Build GRCh38
rs758893105 Chr.11: 72221154-72221154 on Build GRCh38
rs776465189 Chr.11: 72221103-72221104 on Build GRCh38
rs748454608 Chr.11: 72221065-72221065 on Build GRCh38
rs772736516 Chr.11: 72221070-72221070 on Build GRCh38
rs753849601 Chr.11: 72221077-72221077 on Build GRCh38
rs770581420 Chr.11: 72221083-72221083 on Build GRCh38
rs761655995 Chr.11: 72221201-72221201 on Build GRCh38
rs775603112 Chr.11: 72221225-72221225 on Build GRCh38
rs766531689 Chr.11: 72221103-72221103 on Build GRCh38
rs755778536 Chr.11: 72221253-72221253 on Build GRCh38
rs770327598 Chr.11: 72221071-72221071 on Build GRCh38
rs758267564 Chr.11: 72221047-72221047 on Build GRCh38
rs756142886 Chr.11: 72221054-72221054 on Build GRCh38
rs550903576 Chr.11: 72221112-72221112 on Build GRCh38
rs374806625 Chr.11: 72221212-72221212 on Build GRCh38
rs750076536 Chr.11: 72221251-72221251 on Build GRCh38
rs765292321 Chr.11: 72221037-72221037 on Build GRCh38
rs780974334 Chr.11: 72221181-72221181 on Build GRCh38
rs775150042 Chr.11: 72221076-72221076 on Build GRCh38
rs112879648 Chr.11: 72221185-72221185 on Build GRCh38
rs2508202 Chr.11: 72221076-72221076 on Build GRCh38
rs753271188 Chr.11: 72221081-72221081 on Build GRCh38
rs373645194 Chr.11: 72221200-72221200 on Build GRCh38
rs765196424 Chr.11: 72221074-72221074 on Build GRCh38
rs1025089511 Chr.11: 72221227-72221227 on Build GRCh38
rs780992606 Chr.11: 72221084-72221084 on Build GRCh38
rs200999000 Chr.11: 72221222-72221222 on Build GRCh38

View Full Product Details