Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42962575-42962846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Gene Symbol
GNMT
Entrez Gene ID
27232
Gene Name
glycine N-methyltransferase
Gene Aliases
HEL-S-182mP
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42960754-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672419 c.339C>T p.I113I Chr.6: 42962766-42962766 on Build GRCh38
6302111 c.342A>G p.E114E Chr.6: 42962769-42962769 on Build GRCh38
1079380 c.392G>A p.G131D Chr.6: 42962819-42962819 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376412328 Chr.6: 42962795-42962795 on Build GRCh38
rs751482570 Chr.6: 42962595-42962595 on Build GRCh38
rs570363037 Chr.6: 42962696-42962696 on Build GRCh38
rs769257752 Chr.6: 42962719-42962719 on Build GRCh38
rs950102375 Chr.6: 42962677-42962677 on Build GRCh38
rs143043221 Chr.6: 42962800-42962800 on Build GRCh38
rs748566577 Chr.6: 42962724-42962724 on Build GRCh38
rs977270384 Chr.6: 42962707-42962707 on Build GRCh38
rs888783198 Chr.6: 42962631-42962631 on Build GRCh38
rs561305071 Chr.6: 42962649-42962649 on Build GRCh38
rs529994595 Chr.6: 42962639-42962639 on Build GRCh38
rs763777365 Chr.6: 42962778-42962778 on Build GRCh38
rs759363288 Chr.6: 42962766-42962766 on Build GRCh38
rs748483425 Chr.6: 42962739-42962740 on Build GRCh38
rs908525566 Chr.6: 42962705-42962705 on Build GRCh38
rs1021391829 Chr.6: 42962650-42962650 on Build GRCh38
rs530437991 Chr.6: 42962662-42962662 on Build GRCh38
rs751256417 Chr.6: 42962794-42962794 on Build GRCh38
rs9381227 Chr.6: 42962673-42962673 on Build GRCh38
rs776738631 Chr.6: 42962765-42962765 on Build GRCh38
rs370786846 Chr.6: 42962767-42962767 on Build GRCh38
rs772126379 Chr.6: 42962730-42962730 on Build GRCh38
rs979343661 Chr.6: 42962621-42962621 on Build GRCh38
rs550187950 Chr.6: 42962692-42962692 on Build GRCh38
rs370454543 Chr.6: 42962817-42962817 on Build GRCh38
rs968416888 Chr.6: 42962694-42962694 on Build GRCh38
rs997019650 Chr.6: 42962741-42962741 on Build GRCh38
rs541524958 Chr.6: 42962601-42962601 on Build GRCh38
rs375296683 Chr.6: 42962774-42962774 on Build GRCh38
rs770984841 Chr.6: 42962744-42962744 on Build GRCh38
rs749978652 Chr.6: 42962810-42962810 on Build GRCh38
rs115749165 Chr.6: 42962721-42962721 on Build GRCh38
rs745660734 Chr.6: 42962732-42962732 on Build GRCh38
rs984054622 Chr.6: 42962791-42962791 on Build GRCh38
rs747297365 Chr.6: 42962734-42962734 on Build GRCh38
rs369254135 Chr.6: 42962607-42962607 on Build GRCh38

View Full Product Details