Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48728315-48728554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB13
Entrez Gene ID
10481
Gene Name
homeobox B13
Gene Aliases
PSGD
UniGene
Hs.66731
Gene Chromosome Location
Chr.17: 48724763-48728749 on Build GRCh38
Transcripts
NM_006361.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6975742 c.157C>A p.L53M Chr.17: 48728437-48728437 on Build GRCh38
3889826 c.63G>A p.A21A Chr.17: 48728531-48728531 on Build GRCh38
390113 c.271C>A p.R91R Chr.17: 48728323-48728323 on Build GRCh38
1384020 c.85G>A p.A29T Chr.17: 48728509-48728509 on Build GRCh38
3889825 c.251G>A p.G84E Chr.17: 48728343-48728343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529392210 Chr.17: 48728341-48728341 on Build GRCh38
rs776203446 Chr.17: 48728375-48728375 on Build GRCh38
rs587780160 Chr.17: 48728481-48728481 on Build GRCh38
rs763353615 Chr.17: 48728376-48728376 on Build GRCh38
rs750621041 Chr.17: 48728385-48728385 on Build GRCh38
rs758166293 Chr.17: 48728407-48728407 on Build GRCh38
rs769323553 Chr.17: 48728527-48728527 on Build GRCh38
rs756970136 Chr.17: 48728509-48728509 on Build GRCh38
rs550726919 Chr.17: 48728488-48728488 on Build GRCh38
rs370934116 Chr.17: 48728386-48728386 on Build GRCh38
rs568967699 Chr.17: 48728412-48728412 on Build GRCh38
rs774872614 Chr.17: 48728453-48728453 on Build GRCh38
rs766443552 Chr.17: 48728389-48728389 on Build GRCh38
rs199813155 Chr.17: 48728406-48728406 on Build GRCh38
rs587780161 Chr.17: 48728395-48728395 on Build GRCh38
rs774579054 Chr.17: 48728379-48728379 on Build GRCh38
rs772484566 Chr.17: 48728532-48728532 on Build GRCh38
rs779780541 Chr.17: 48728417-48728417 on Build GRCh38
rs760874697 Chr.17: 48728479-48728479 on Build GRCh38
rs768670673 Chr.17: 48728384-48728384 on Build GRCh38
rs778843798 Chr.17: 48728398-48728398 on Build GRCh38
rs539706443 Chr.17: 48728530-48728530 on Build GRCh38
rs762197066 Chr.17: 48728383-48728383 on Build GRCh38
rs866159706 Chr.17: 48728463-48728463 on Build GRCh38
rs751338230 Chr.17: 48728505-48728505 on Build GRCh38
rs754872559 Chr.17: 48728336-48728336 on Build GRCh38
rs368505949 Chr.17: 48728531-48728531 on Build GRCh38
rs748333323 Chr.17: 48728454-48728454 on Build GRCh38
rs769145488 Chr.17: 48728378-48728378 on Build GRCh38
rs924383022 Chr.17: 48728360-48728360 on Build GRCh38
rs745306184 Chr.17: 48728425-48728425 on Build GRCh38
rs958681537 Chr.17: 48728358-48728358 on Build GRCh38
rs587780165 Chr.17: 48728520-48728520 on Build GRCh38
rs541519790 Chr.17: 48728411-48728411 on Build GRCh38
rs752661579 Chr.17: 48728498-48728498 on Build GRCh38
rs570672959 Chr.17: 48728495-48728495 on Build GRCh38
rs770545259 Chr.17: 48728363-48728363 on Build GRCh38
rs772962401 Chr.17: 48728355-48728355 on Build GRCh38
rs758169931 Chr.17: 48728502-48728502 on Build GRCh38
rs746542615 Chr.17: 48728362-48728362 on Build GRCh38
rs79344505 Chr.17: 48728497-48728497 on Build GRCh38
rs773491778 Chr.17: 48728478-48728478 on Build GRCh38
rs778563157 Chr.17: 48728340-48728340 on Build GRCh38
rs754885900 Chr.17: 48728396-48728396 on Build GRCh38
rs780859632 Chr.17: 48728423-48728423 on Build GRCh38
rs770620686 Chr.17: 48728418-48728418 on Build GRCh38
rs745392080 Chr.17: 48728526-48728526 on Build GRCh38
rs561048036 Chr.17: 48728504-48728504 on Build GRCh38
rs138213197 Chr.17: 48728343-48728343 on Build GRCh38
rs769074510 Chr.17: 48728434-48728434 on Build GRCh38
rs199799743 Chr.17: 48728472-48728472 on Build GRCh38
rs11551892 Chr.17: 48728449-48728449 on Build GRCh38
rs780947625 Chr.17: 48728524-48728524 on Build GRCh38
rs1053531133 Chr.17: 48728519-48728519 on Build GRCh38

View Full Product Details