Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1092181-1092430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
UniGene
Hs.20961
Gene Chromosome Location
Chr.7: 1086807-1093815 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3698105 c.586G>A p.V196M Chr.7: 1092314-1092314 on Build GRCh38
1084160 c.672C>T p.I224I Chr.7: 1092400-1092400 on Build GRCh38
1084159 c.524G>A p.R175Q Chr.7: 1092252-1092252 on Build GRCh38
6235654 c.632T>C p.V211A Chr.7: 1092360-1092360 on Build GRCh38
4687793 c.519C>T p.H173H Chr.7: 1092247-1092247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376219774 Chr.7: 1092223-1092223 on Build GRCh38
rs368423693 Chr.7: 1092312-1092312 on Build GRCh38
rs771510444 Chr.7: 1092401-1092401 on Build GRCh38
rs768379637 Chr.7: 1092394-1092394 on Build GRCh38
rs199951210 Chr.7: 1092354-1092354 on Build GRCh38
rs376117751 Chr.7: 1092284-1092284 on Build GRCh38
rs750810991 Chr.7: 1092200-1092200 on Build GRCh38
rs778619173 Chr.7: 1092400-1092400 on Build GRCh38
rs752930799 Chr.7: 1092370-1092370 on Build GRCh38
rs748855645 Chr.7: 1092320-1092320 on Build GRCh38
rs756464399 Chr.7: 1092277-1092277 on Build GRCh38
rs147112257 Chr.7: 1092294-1092294 on Build GRCh38
rs771633136 Chr.7: 1092334-1092334 on Build GRCh38
rs749090970 Chr.7: 1092389-1092389 on Build GRCh38
rs148096895 Chr.7: 1092310-1092310 on Build GRCh38
rs766783283 Chr.7: 1092283-1092283 on Build GRCh38
rs780000717 Chr.7: 1092313-1092313 on Build GRCh38
rs367650982 Chr.7: 1092212-1092212 on Build GRCh38
rs776517088 Chr.7: 1092239-1092239 on Build GRCh38
rs199736734 Chr.7: 1092380-1092380 on Build GRCh38
rs746854600 Chr.7: 1092226-1092226 on Build GRCh38
rs748280302 Chr.7: 1092295-1092295 on Build GRCh38
rs754995478 Chr.7: 1092222-1092222 on Build GRCh38
rs759628974 Chr.7: 1092363-1092363 on Build GRCh38
rs768049762 Chr.7: 1092263-1092263 on Build GRCh38
rs773691766 Chr.7: 1092339-1092339 on Build GRCh38
rs775268794 Chr.7: 1092248-1092248 on Build GRCh38
rs771448862 Chr.7: 1092410-1092410 on Build GRCh38
rs763449832 Chr.7: 1092252-1092252 on Build GRCh38
rs770930808 Chr.7: 1092233-1092233 on Build GRCh38
rs754230934 Chr.7: 1092362-1092362 on Build GRCh38
rs201825687 Chr.7: 1092220-1092220 on Build GRCh38
rs375858677 Chr.7: 1092332-1092332 on Build GRCh38
rs750319978 Chr.7: 1092382-1092382 on Build GRCh38
rs762179730 Chr.7: 1092261-1092261 on Build GRCh38
rs766662470 Chr.7: 1092359-1092359 on Build GRCh38
rs772711029 Chr.7: 1092406-1092406 on Build GRCh38
rs370862534 Chr.7: 1092247-1092247 on Build GRCh38
rs778014567 Chr.7: 1092381-1092381 on Build GRCh38
rs768379201 Chr.7: 1092324-1092324 on Build GRCh38
rs141830499 Chr.7: 1092311-1092311 on Build GRCh38
rs780215746 Chr.7: 1092216-1092216 on Build GRCh38
rs758456215 Chr.7: 1092301-1092301 on Build GRCh38
rs150646955 Chr.7: 1092387-1092387 on Build GRCh38
rs372712083 Chr.7: 1092331-1092331 on Build GRCh38
rs1025216784 Chr.7: 1092379-1092379 on Build GRCh38
rs200198507 Chr.7: 1092265-1092265 on Build GRCh38
rs370109208 Chr.7: 1092259-1092259 on Build GRCh38
rs756154497 Chr.7: 1092384-1092384 on Build GRCh38
rs761181037 Chr.7: 1092355-1092355 on Build GRCh38
rs201528297 Chr.7: 1092251-1092251 on Build GRCh38
rs139741139 Chr.7: 1092409-1092409 on Build GRCh38
rs765474183 Chr.7: 1092367-1092367 on Build GRCh38
rs745627977 Chr.7: 1092244-1092244 on Build GRCh38

View Full Product Details