Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244705419-244705650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DESI2
Entrez Gene ID
51029
Gene Name
desumoylating isopeptidase 2
Gene Aliases
C1orf121, CGI-146, DESI, DESI1, DeSI-2, FAM152A, PNAS-4, PPPDE1
UniGene
Hs.498317
Gene Chromosome Location
Chr.1: 244653048-244709034 on Build GRCh38
Transcripts
NM_001297746.1 NM_016076.4 NR_123735.1 NR_123736.1 XM_011544203.2 XM_011544205.2 XM_011544206.2 XM_011544207.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735922 c.424C>T p.Q142* Chr.1: 244705628-244705628 on Build GRCh38
3789754 c.355C>G p.L119V Chr.1: 244705559-244705559 on Build GRCh38
171015 c.355C>A p.L119I Chr.1: 244705559-244705559 on Build GRCh38
4545611 c.381G>A p.W127* Chr.1: 244705585-244705585 on Build GRCh38
1663658 c.371T>A p.I124N Chr.1: 244705575-244705575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953653365 Chr.1: 244705469-244705469 on Build GRCh38
rs763270053 Chr.1: 244705612-244705612 on Build GRCh38
rs530003762 Chr.1: 244705633-244705633 on Build GRCh38
rs756755760 Chr.1: 244705506-244705506 on Build GRCh38
rs894740503 Chr.1: 244705448-244705448 on Build GRCh38
rs778968742 Chr.1: 244705479-244705479 on Build GRCh38
rs776536832 Chr.1: 244705550-244705550 on Build GRCh38
rs781187398 Chr.1: 244705542-244705543 on Build GRCh38
rs1042778089 Chr.1: 244705478-244705478 on Build GRCh38
rs537211255 Chr.1: 244705463-244705463 on Build GRCh38
rs370829407 Chr.1: 244705552-244705552 on Build GRCh38
rs998607094 Chr.1: 244705504-244705504 on Build GRCh38
rs957053470 Chr.1: 244705442-244705442 on Build GRCh38
rs757296615 Chr.1: 244705445-244705445 on Build GRCh38
rs778513614 Chr.1: 244705507-244705507 on Build GRCh38
rs745579344 Chr.1: 244705518-244705518 on Build GRCh38
rs554315585 Chr.1: 244705472-244705472 on Build GRCh38
rs13270 Chr.1: 244705602-244705602 on Build GRCh38
rs894368417 Chr.1: 244705567-244705567 on Build GRCh38
rs902815761 Chr.1: 244705486-244705486 on Build GRCh38
rs1019351644 Chr.1: 244705530-244705530 on Build GRCh38
rs758759055 Chr.1: 244705519-244705519 on Build GRCh38
rs770050624 Chr.1: 244705582-244705582 on Build GRCh38
rs200355784 Chr.1: 244705504-244705504 on Build GRCh38
rs747416996 Chr.1: 244705529-244705529 on Build GRCh38
rs986081371 Chr.1: 244705470-244705470 on Build GRCh38
rs551183369 Chr.1: 244705464-244705464 on Build GRCh38
rs769231480 Chr.1: 244705533-244705533 on Build GRCh38
rs754813073 Chr.1: 244705539-244705540 on Build GRCh38
rs773561417 Chr.1: 244705606-244705606 on Build GRCh38
rs915475137 Chr.1: 244705467-244705467 on Build GRCh38

View Full Product Details