Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41216128-41216353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHADL
Entrez Gene ID
150356
Gene Name
chondroadherin like
Gene Aliases
SLRR4B
UniGene
Hs.344488
Gene Chromosome Location
Chr.22: 41209101-41241773 on Build GRCh38
Transcripts
XM_005261370.3 XM_011529932.2 XM_011529934.2 NM_031488.4 XM_011530420.2 XM_011530421.2 XM_017028976.1 XM_017028977.1 XM_017028978.1 XM_017028979.1 XM_017028980.1

Gene Symbol
L3MBTL2
Entrez Gene ID
83746
Gene Name
L3MBTL2 polycomb repressive complex 1 subunit
Gene Aliases
H-l(3)mbt-l, L3MBT
UniGene
Hs.517641
Gene Chromosome Location
Chr.22: 41205192-41231271 on Build GRCh38
Transcripts
XM_005261370.3 XM_011529932.2 XM_011529934.2 NM_031488.4 XM_011530420.2 XM_011530421.2 XM_017028976.1 XM_017028977.1 XM_017028978.1 XM_017028979.1 XM_017028980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7279682 c.412delA p.A140fs*46 Chr.22: 41216154-41216154 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776319944 Chr.22: 41216270-41216272 on Build GRCh38
rs763485284 Chr.22: 41216231-41216231 on Build GRCh38
rs766846536 Chr.22: 41216232-41216232 on Build GRCh38
rs373234786 Chr.22: 41216297-41216297 on Build GRCh38
rs778603987 Chr.22: 41216262-41216262 on Build GRCh38
rs763592335 Chr.22: 41216246-41216246 on Build GRCh38
rs201167800 Chr.22: 41216291-41216291 on Build GRCh38
rs755116539 Chr.22: 41216283-41216283 on Build GRCh38
rs554400330 Chr.22: 41216260-41216260 on Build GRCh38
rs577165731 Chr.22: 41216293-41216293 on Build GRCh38
rs1004694318 Chr.22: 41216305-41216305 on Build GRCh38
rs530184410 Chr.22: 41216300-41216300 on Build GRCh38
rs747153740 Chr.22: 41216278-41216278 on Build GRCh38
rs781278136 Chr.22: 41216286-41216286 on Build GRCh38
rs1019206797 Chr.22: 41216323-41216323 on Build GRCh38
rs544192051 Chr.22: 41216324-41216324 on Build GRCh38
rs759977115 Chr.22: 41216304-41216304 on Build GRCh38
rs1055920501 Chr.22: 41216171-41216171 on Build GRCh38
rs559679859 Chr.22: 41216298-41216298 on Build GRCh38
rs201967099 Chr.22: 41216271-41216271 on Build GRCh38
rs145875010 Chr.22: 41216261-41216261 on Build GRCh38
rs769058510 Chr.22: 41216187-41216187 on Build GRCh38
rs750213400 Chr.22: 41216269-41216269 on Build GRCh38
rs752257584 Chr.22: 41216233-41216233 on Build GRCh38
rs771492070 Chr.22: 41216299-41216299 on Build GRCh38
rs898480790 Chr.22: 41216217-41216217 on Build GRCh38
rs774841393 Chr.22: 41216301-41216301 on Build GRCh38
rs770021384 Chr.22: 41216292-41216292 on Build GRCh38
rs888873642 Chr.22: 41216320-41216320 on Build GRCh38
rs762031971 Chr.22: 41216222-41216222 on Build GRCh38
rs776714489 Chr.22: 41216216-41216216 on Build GRCh38
rs377308699 Chr.22: 41216224-41216224 on Build GRCh38
rs760169500 Chr.22: 41216243-41216243 on Build GRCh38
rs780322228 Chr.22: 41216174-41216174 on Build GRCh38
rs773579150 Chr.22: 41216230-41216230 on Build GRCh38
rs536318588 Chr.22: 41216150-41216150 on Build GRCh38

View Full Product Details