Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74844417-74844642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2C
Entrez Gene ID
2905
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Gene Aliases
GluN2C, NMDAR2C, NR2C
UniGene
Hs.436980
Gene Chromosome Location
Chr.17: 74842021-74860843 on Build GRCh38
Transcripts
NM_000835.4 NM_001278553.1 NR_103735.1 XM_006721845.3 XM_006721846.3 XM_011524686.2 XM_011524687.2 XM_011524688.2 XM_011524689.2 XM_011524692.2 XM_017024544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707296 c.2384C>T p.S795L Chr.17: 74844475-74844475 on Build GRCh38
984126 c.2433C>T p.D811D Chr.17: 74844426-74844426 on Build GRCh38
3820511 c.2391C>T p.I797I Chr.17: 74844468-74844468 on Build GRCh38
437338 c.2387G>A p.G796E Chr.17: 74844472-74844472 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs936937481 Chr.17: 74844559-74844559 on Build GRCh38
rs775241120 Chr.17: 74844493-74844493 on Build GRCh38
rs763568207 Chr.17: 74844550-74844550 on Build GRCh38
rs559425028 Chr.17: 74844603-74844603 on Build GRCh38
rs752303974 Chr.17: 74844440-74844440 on Build GRCh38
rs927144479 Chr.17: 74844549-74844549 on Build GRCh38
rs755661188 Chr.17: 74844547-74844547 on Build GRCh38
rs747627917 Chr.17: 74844527-74844527 on Build GRCh38
rs138088491 Chr.17: 74844487-74844487 on Build GRCh38
rs780908833 Chr.17: 74844517-74844517 on Build GRCh38
rs753393714 Chr.17: 74844560-74844560 on Build GRCh38
rs751191029 Chr.17: 74844451-74844453 on Build GRCh38
rs754674133 Chr.17: 74844497-74844498 on Build GRCh38
rs576642341 Chr.17: 74844450-74844450 on Build GRCh38
rs925515965 Chr.17: 74844497-74844497 on Build GRCh38
rs753075824 Chr.17: 74844544-74844544 on Build GRCh38
rs759874906 Chr.17: 74844529-74844529 on Build GRCh38
rs751964847 Chr.17: 74844528-74844528 on Build GRCh38
rs965016994 Chr.17: 74844453-74844453 on Build GRCh38
rs369028851 Chr.17: 74844441-74844441 on Build GRCh38
rs766587072 Chr.17: 74844526-74844526 on Build GRCh38
rs768097005 Chr.17: 74844534-74844534 on Build GRCh38
rs769698308 Chr.17: 74844504-74844504 on Build GRCh38
rs377010808 Chr.17: 74844589-74844589 on Build GRCh38
rs983970064 Chr.17: 74844494-74844494 on Build GRCh38
rs745627652 Chr.17: 74844486-74844486 on Build GRCh38
rs201650252 Chr.17: 74844575-74844577 on Build GRCh38
rs763220355 Chr.17: 74844519-74844519 on Build GRCh38
rs889431032 Chr.17: 74844588-74844588 on Build GRCh38
rs773497242 Chr.17: 74844517-74844517 on Build GRCh38
rs777483624 Chr.17: 74844446-74844446 on Build GRCh38
rs761985013 Chr.17: 74844501-74844501 on Build GRCh38
rs374337919 Chr.17: 74844462-74844462 on Build GRCh38
rs113050737 Chr.17: 74844474-74844474 on Build GRCh38
rs991284300 Chr.17: 74844597-74844597 on Build GRCh38

View Full Product Details