Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109974070-109974329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV1
Entrez Gene ID
27012
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
HNKA, KCNB3, KV2.3, KV8.1
UniGene
Hs.13285
Gene Chromosome Location
Chr.8: 109963645-109975847 on Build GRCh38
Transcripts
NM_014379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533146 c.94G>A p.G32R Chr.8: 109974295-109974295 on Build GRCh38
4651618 c.111C>T p.L37L Chr.8: 109974278-109974278 on Build GRCh38
453869 c.227G>T p.R76L Chr.8: 109974162-109974162 on Build GRCh38
5445145 c.115G>T p.D39Y Chr.8: 109974274-109974274 on Build GRCh38
3884764 c.309C>T p.F103F Chr.8: 109974080-109974080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961500345 Chr.8: 109974290-109974290 on Build GRCh38
rs749513053 Chr.8: 109974233-109974233 on Build GRCh38
rs1030119362 Chr.8: 109974155-109974155 on Build GRCh38
rs200185762 Chr.8: 109974165-109974165 on Build GRCh38
rs924158633 Chr.8: 109974215-109974215 on Build GRCh38
rs1046900437 Chr.8: 109974280-109974280 on Build GRCh38
rs775731785 Chr.8: 109974300-109974300 on Build GRCh38
rs954505973 Chr.8: 109974168-109974168 on Build GRCh38
rs750493625 Chr.8: 109974200-109974200 on Build GRCh38
rs764901299 Chr.8: 109974101-109974101 on Build GRCh38
rs936911224 Chr.8: 109974256-109974256 on Build GRCh38
rs752311853 Chr.8: 109974113-109974113 on Build GRCh38
rs140779438 Chr.8: 109974230-109974230 on Build GRCh38
rs559355620 Chr.8: 109974143-109974143 on Build GRCh38
rs977778850 Chr.8: 109974203-109974203 on Build GRCh38
rs984370595 Chr.8: 109974277-109974277 on Build GRCh38
rs866849804 Chr.8: 109974108-109974108 on Build GRCh38
rs916729786 Chr.8: 109974275-109974275 on Build GRCh38
rs751534416 Chr.8: 109974166-109974166 on Build GRCh38
rs1018981507 Chr.8: 109974161-109974161 on Build GRCh38
rs1000789197 Chr.8: 109974091-109974091 on Build GRCh38
rs371539902 Chr.8: 109974170-109974170 on Build GRCh38
rs751459891 Chr.8: 109974092-109974093 on Build GRCh38
rs769794827 Chr.8: 109974263-109974263 on Build GRCh38
rs757208349 Chr.8: 109974167-109974167 on Build GRCh38
rs148023486 Chr.8: 109974158-109974158 on Build GRCh38
rs974890055 Chr.8: 109974139-109974139 on Build GRCh38
rs377263756 Chr.8: 109974278-109974278 on Build GRCh38
rs763831675 Chr.8: 109974144-109974144 on Build GRCh38
rs112430047 Chr.8: 109974159-109974159 on Build GRCh38
rs867957211 Chr.8: 109974112-109974112 on Build GRCh38
rs992612607 Chr.8: 109974308-109974308 on Build GRCh38
rs769061477 Chr.8: 109974257-109974257 on Build GRCh38
rs779307333 Chr.8: 109974274-109974274 on Build GRCh38
rs756173601 Chr.8: 109974224-109974224 on Build GRCh38
rs891882060 Chr.8: 109974145-109974145 on Build GRCh38

View Full Product Details