Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131117713-131117966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIF1L
Entrez Gene ID
83543
Gene Name
allograft inflammatory factor 1 like
Gene Aliases
C9orf58, IBA2
UniGene
Hs.4944
Gene Chromosome Location
Chr.9: 131096476-131123152 on Build GRCh38
Transcripts
NM_001185095.1 NM_001185096.1 NM_031426.3 NR_033701.1 XM_017015177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5923832 c.389C>T p.S130F Chr.9: 131117864-131117864 on Build GRCh38
6472986 c.303C>T p.D101D Chr.9: 131117856-131117856 on Build GRCh38
1244705 c.292G>A p.G98R Chr.9: 131117845-131117845 on Build GRCh38
5991453 c.282C>G p.D94E Chr.9: 131117757-131117757 on Build GRCh38
1244704 c.370G>A p.G124R Chr.9: 131117845-131117845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374128627 Chr.9: 131117748-131117748 on Build GRCh38
rs751087808 Chr.9: 131117822-131117822 on Build GRCh38
rs767468057 Chr.9: 131117835-131117835 on Build GRCh38
rs912197577 Chr.9: 131117898-131117898 on Build GRCh38
rs917168943 Chr.9: 131117877-131117877 on Build GRCh38
rs930919807 Chr.9: 131117907-131117907 on Build GRCh38
rs756703547 Chr.9: 131117831-131117831 on Build GRCh38
rs34166734 Chr.9: 131117904-131117904 on Build GRCh38
rs140648753 Chr.9: 131117854-131117854 on Build GRCh38
rs778945229 Chr.9: 131117889-131117889 on Build GRCh38
rs979274722 Chr.9: 131117833-131117833 on Build GRCh38
rs774483085 Chr.9: 131117787-131117787 on Build GRCh38
rs139271942 Chr.9: 131117931-131117931 on Build GRCh38
rs151301044 Chr.9: 131117796-131117796 on Build GRCh38
rs146631291 Chr.9: 131117918-131117918 on Build GRCh38
rs781045219 Chr.9: 131117752-131117752 on Build GRCh38
rs749537634 Chr.9: 131117870-131117870 on Build GRCh38
rs773805004 Chr.9: 131117800-131117800 on Build GRCh38
rs750325550 Chr.9: 131117838-131117838 on Build GRCh38
rs755231326 Chr.9: 131117883-131117883 on Build GRCh38
rs548343527 Chr.9: 131117811-131117811 on Build GRCh38
rs779882634 Chr.9: 131117869-131117869 on Build GRCh38
rs755859603 Chr.9: 131117764-131117764 on Build GRCh38
rs747498844 Chr.9: 131117905-131117905 on Build GRCh38
rs757889461 Chr.9: 131117742-131117742 on Build GRCh38
rs777053012 Chr.9: 131117919-131117919 on Build GRCh38
rs775055463 Chr.9: 131117807-131117807 on Build GRCh38
rs768672035 Chr.9: 131117780-131117780 on Build GRCh38
rs879392854 Chr.9: 131117900-131117900 on Build GRCh38
rs759755280 Chr.9: 131117921-131117921 on Build GRCh38
rs772239555 Chr.9: 131117903-131117903 on Build GRCh38
rs761153815 Chr.9: 131117801-131117801 on Build GRCh38
rs905399625 Chr.9: 131117934-131117934 on Build GRCh38
rs755681970 Chr.9: 131117852-131117852 on Build GRCh38
rs112041142 Chr.9: 131117745-131117746 on Build GRCh38
rs768372779 Chr.9: 131117923-131117923 on Build GRCh38
rs749501933 Chr.9: 131117775-131117775 on Build GRCh38
rs536561486 Chr.9: 131117899-131117899 on Build GRCh38
rs751513279 Chr.9: 131117749-131117749 on Build GRCh38
rs747327267 Chr.9: 131117738-131117738 on Build GRCh38
rs757185426 Chr.9: 131117750-131117750 on Build GRCh38
rs369690763 Chr.9: 131117848-131117848 on Build GRCh38
rs748107649 Chr.9: 131117789-131117789 on Build GRCh38
rs780290936 Chr.9: 131117774-131117774 on Build GRCh38
rs986518231 Chr.9: 131117856-131117856 on Build GRCh38
rs766820239 Chr.9: 131117805-131117805 on Build GRCh38
rs745679588 Chr.9: 131117756-131117756 on Build GRCh38
rs766987529 Chr.9: 131117933-131117933 on Build GRCh38

View Full Product Details