Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101624934-101625205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX3
Entrez Gene ID
51566
Gene Name
armadillo repeat containing, X-linked 3
Gene Aliases
ALEX3, GASP6, dJ545K15.2
UniGene
Hs.592225
Gene Chromosome Location
Chr.X: 101623127-101631910 on Build GRCh38
Transcripts
NM_016607.3 NM_177947.2 NM_177948.2 XM_005262141.2

SNP Detail

SNP ID Chromosome Location
rs782415759 Chr.X: 101625125-101625125 on Build GRCh38
rs782741127 Chr.X: 101625178-101625178 on Build GRCh38
rs782622259 Chr.X: 101625143-101625143 on Build GRCh38
rs200640542 Chr.X: 101624960-101624960 on Build GRCh38
rs895750150 Chr.X: 101625134-101625134 on Build GRCh38
rs782394011 Chr.X: 101625031-101625031 on Build GRCh38
rs782282127 Chr.X: 101625025-101625025 on Build GRCh38
rs782110371 Chr.X: 101625177-101625177 on Build GRCh38
rs920405923 Chr.X: 101624976-101624976 on Build GRCh38
rs782455924 Chr.X: 101624968-101624968 on Build GRCh38
rs372310822 Chr.X: 101625036-101625036 on Build GRCh38
rs782061543 Chr.X: 101625153-101625153 on Build GRCh38
rs782347784 Chr.X: 101625056-101625056 on Build GRCh38
rs781998690 Chr.X: 101625155-101625155 on Build GRCh38
rs138397719 Chr.X: 101625115-101625115 on Build GRCh38
rs782636138 Chr.X: 101625032-101625032 on Build GRCh38
rs200151007 Chr.X: 101624990-101624990 on Build GRCh38
rs782644531 Chr.X: 101625113-101625113 on Build GRCh38

View Full Product Details