Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44048005-44048246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCSL
Entrez Gene ID
390110
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Gene Aliases
-
UniGene
Hs.558851
Gene Chromosome Location
Chr.11: 43920987-44060667 on Build GRCh38
Transcripts
NM_001031854.2 XM_017017714.1 XM_017017715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7305413 c.153G>T p.Q51H Chr.11: 44048189-44048189 on Build GRCh38
4392324 c.177G>A p.R59R Chr.11: 44048213-44048213 on Build GRCh38
6132862 c.40C>G p.Q14E Chr.11: 44048076-44048076 on Build GRCh38
7314566 c.53G>T p.R18L Chr.11: 44048089-44048089 on Build GRCh38
3724744 c.64G>C p.D22H Chr.11: 44048100-44048100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763059170 Chr.11: 44048074-44048074 on Build GRCh38
rs376520618 Chr.11: 44048197-44048197 on Build GRCh38
rs750151692 Chr.11: 44048098-44048099 on Build GRCh38
rs776658808 Chr.11: 44048176-44048176 on Build GRCh38
rs750164034 Chr.11: 44048144-44048144 on Build GRCh38
rs1026879463 Chr.11: 44048116-44048116 on Build GRCh38
rs995127281 Chr.11: 44048113-44048113 on Build GRCh38
rs756899266 Chr.11: 44048123-44048123 on Build GRCh38
rs3094398 Chr.11: 44048039-44048039 on Build GRCh38
rs757125268 Chr.11: 44048037-44048037 on Build GRCh38
rs754482700 Chr.11: 44048117-44048118 on Build GRCh38
rs759303797 Chr.11: 44048210-44048210 on Build GRCh38
rs200626992 Chr.11: 44048184-44048184 on Build GRCh38
rs779631822 Chr.11: 44048108-44048111 on Build GRCh38
rs768077997 Chr.11: 44048092-44048092 on Build GRCh38
rs998442475 Chr.11: 44048043-44048043 on Build GRCh38
rs558611369 Chr.11: 44048035-44048035 on Build GRCh38
rs755914763 Chr.11: 44048102-44048105 on Build GRCh38
rs752150289 Chr.11: 44048093-44048093 on Build GRCh38
rs779207509 Chr.11: 44048046-44048046 on Build GRCh38
rs984830077 Chr.11: 44048141-44048141 on Build GRCh38
rs761472684 Chr.11: 44048202-44048202 on Build GRCh38
rs748197340 Chr.11: 44048168-44048168 on Build GRCh38
rs753521392 Chr.11: 44048114-44048117 on Build GRCh38
rs200325579 Chr.11: 44048179-44048179 on Build GRCh38
rs374627783 Chr.11: 44048225-44048225 on Build GRCh38
rs561571804 Chr.11: 44048217-44048217 on Build GRCh38
rs201252067 Chr.11: 44048031-44048031 on Build GRCh38
rs774851363 Chr.11: 44048089-44048089 on Build GRCh38
rs758137938 Chr.11: 44048218-44048218 on Build GRCh38
rs769938441 Chr.11: 44048067-44048067 on Build GRCh38
rs748246175 Chr.11: 44048108-44048108 on Build GRCh38
rs748194606 Chr.11: 44048066-44048066 on Build GRCh38
rs377516400 Chr.11: 44048159-44048159 on Build GRCh38
rs374220502 Chr.11: 44048158-44048158 on Build GRCh38
rs760178254 Chr.11: 44048090-44048090 on Build GRCh38
rs763490870 Chr.11: 44048106-44048106 on Build GRCh38
rs899402486 Chr.11: 44048088-44048088 on Build GRCh38
rs776707991 Chr.11: 44048060-44048060 on Build GRCh38
rs370566395 Chr.11: 44048153-44048153 on Build GRCh38
rs750251835 Chr.11: 44048213-44048213 on Build GRCh38
rs377118963 Chr.11: 44048134-44048134 on Build GRCh38
rs909188682 Chr.11: 44048148-44048148 on Build GRCh38
rs760768129 Chr.11: 44048163-44048163 on Build GRCh38
rs766885931 Chr.11: 44048086-44048086 on Build GRCh38
rs372620308 Chr.11: 44048068-44048068 on Build GRCh38
rs755733351 Chr.11: 44048030-44048030 on Build GRCh38
rs759910901 Chr.11: 44048192-44048192 on Build GRCh38
rs11037840 Chr.11: 44048169-44048169 on Build GRCh38
rs190500805 Chr.11: 44048198-44048198 on Build GRCh38
rs771264543 Chr.11: 44048180-44048180 on Build GRCh38
rs934380761 Chr.11: 44048032-44048032 on Build GRCh38
rs1003129570 Chr.11: 44048100-44048100 on Build GRCh38
rs760185887 Chr.11: 44048094-44048094 on Build GRCh38
rs772730631 Chr.11: 44048193-44048193 on Build GRCh38
rs746240408 Chr.11: 44048099-44048099 on Build GRCh38
rs370567159 Chr.11: 44048047-44048047 on Build GRCh38

View Full Product Details