Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1451851-1452112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3C
Entrez Gene ID
219293
Gene Name
ATPase family, AAA domain containing 3C
Gene Aliases
-
UniGene
Hs.724767
Gene Chromosome Location
Chr.1: 1449689-1470158 on Build GRCh38
Transcripts
NM_001039211.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7108308 c.95T>C p.L32S Chr.1: 1452065-1452065 on Build GRCh38
7356768 c.91G>A p.D31N Chr.1: 1452061-1452061 on Build GRCh38
6019428 c.106G>C p.E36Q Chr.1: 1452076-1452076 on Build GRCh38
5968780 c.135C>T p.T45T Chr.1: 1452105-1452105 on Build GRCh38
7144174 c.130C>G p.Q44E Chr.1: 1452100-1452100 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563980453 Chr.1: 1452040-1452040 on Build GRCh38
rs894032869 Chr.1: 1451901-1451901 on Build GRCh38
rs1781142 Chr.1: 1451913-1451913 on Build GRCh38
rs1024573545 Chr.1: 1451935-1451935 on Build GRCh38
rs762275330 Chr.1: 1452057-1452058 on Build GRCh38
rs919792061 Chr.1: 1451940-1451940 on Build GRCh38
rs564349232 Chr.1: 1451953-1451953 on Build GRCh38
rs146946254 Chr.1: 1452067-1452067 on Build GRCh38
rs954539077 Chr.1: 1451972-1451972 on Build GRCh38
rs745945408 Chr.1: 1452061-1452061 on Build GRCh38
rs539395811 Chr.1: 1451886-1451886 on Build GRCh38
rs748157292 Chr.1: 1452073-1452073 on Build GRCh38
rs756359829 Chr.1: 1452020-1452021 on Build GRCh38
rs765747210 Chr.1: 1452033-1452033 on Build GRCh38
rs951484742 Chr.1: 1451948-1451948 on Build GRCh38
rs1025721752 Chr.1: 1451945-1451945 on Build GRCh38
rs768658521 Chr.1: 1452061-1452062 on Build GRCh38
rs769047453 Chr.1: 1451896-1451896 on Build GRCh38
rs774306838 Chr.1: 1452065-1452066 on Build GRCh38
rs889826039 Chr.1: 1451881-1451881 on Build GRCh38
rs34358987 Chr.1: 1452034-1452035 on Build GRCh38
rs376115555 Chr.1: 1451914-1451914 on Build GRCh38
rs753269251 Chr.1: 1452035-1452035 on Build GRCh38
rs1005529390 Chr.1: 1451951-1451951 on Build GRCh38
rs12032431 Chr.1: 1452075-1452075 on Build GRCh38
rs113840210 Chr.1: 1451902-1451902 on Build GRCh38
rs185554649 Chr.1: 1451993-1451993 on Build GRCh38
rs12032405 Chr.1: 1452045-1452045 on Build GRCh38
rs781484435 Chr.1: 1452032-1452032 on Build GRCh38
rs372727489 Chr.1: 1452085-1452085 on Build GRCh38
rs904730367 Chr.1: 1452000-1452000 on Build GRCh38
rs946144240 Chr.1: 1451973-1451973 on Build GRCh38
rs961133150 Chr.1: 1451938-1451938 on Build GRCh38
rs911023312 Chr.1: 1452071-1452071 on Build GRCh38
rs747365372 Chr.1: 1452043-1452043 on Build GRCh38
rs112072883 Chr.1: 1452052-1452052 on Build GRCh38
rs532640633 Chr.1: 1451969-1451969 on Build GRCh38
rs111237704 Chr.1: 1451885-1451885 on Build GRCh38
rs112911422 Chr.1: 1451873-1451873 on Build GRCh38
rs781422464 Chr.1: 1452057-1452057 on Build GRCh38
rs544624392 Chr.1: 1452068-1452068 on Build GRCh38
rs753321251 Chr.1: 1452021-1452021 on Build GRCh38
rs111805964 Chr.1: 1451939-1451939 on Build GRCh38
rs766021483 Chr.1: 1452006-1452006 on Build GRCh38
rs778682621 Chr.1: 1451916-1451916 on Build GRCh38
rs562186185 Chr.1: 1451917-1451917 on Build GRCh38
rs779661490 Chr.1: 1451997-1451997 on Build GRCh38
rs943923979 Chr.1: 1451958-1451958 on Build GRCh38
rs1021825726 Chr.1: 1451897-1451897 on Build GRCh38
rs760480398 Chr.1: 1452087-1452087 on Build GRCh38
rs541398798 Chr.1: 1451952-1451952 on Build GRCh38
rs1034900866 Chr.1: 1452008-1452008 on Build GRCh38
rs368507723 Chr.1: 1451877-1451877 on Build GRCh38
rs963293770 Chr.1: 1451966-1451966 on Build GRCh38
rs754595153 Chr.1: 1451957-1451957 on Build GRCh38
rs1781143 Chr.1: 1451968-1451968 on Build GRCh38
rs112013420 Chr.1: 1451882-1451882 on Build GRCh38
rs748033393 Chr.1: 1452074-1452076 on Build GRCh38
rs772115111 Chr.1: 1452084-1452084 on Build GRCh38
rs931250896 Chr.1: 1451876-1451876 on Build GRCh38
rs961020331 Chr.1: 1452041-1452041 on Build GRCh38
rs746379288 Chr.1: 1452037-1452037 on Build GRCh38
rs760291516 Chr.1: 1452001-1452001 on Build GRCh38
rs931157250 Chr.1: 1451944-1451944 on Build GRCh38
rs1001717365 Chr.1: 1452003-1452003 on Build GRCh38
rs780310897 Chr.1: 1452035-1452037 on Build GRCh38

View Full Product Details