Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4167660-4167925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
UniGene
Hs.696087
Gene Chromosome Location
Chr.17: 4163371-4264026 on Build GRCh38
Transcripts
NM_001257999.1 NM_016376.3 NR_047571.1 XM_005256679.3 XM_011523926.1 XM_011523928.2 XM_011523933.1 XM_017024733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
335824 c.3479G>C p.C1160S Chr.17: 4167813-4167813 on Build GRCh38
3188920 c.3431C>T p.P1144L Chr.17: 4167861-4167861 on Build GRCh38
3712317 c.3474C>A p.N1158K Chr.17: 4167818-4167818 on Build GRCh38
5778129 c.3430C>G p.P1144A Chr.17: 4167862-4167862 on Build GRCh38
6523168 c.3527G>T p.C1176F Chr.17: 4167888-4167888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754412631 Chr.17: 4167775-4167778 on Build GRCh38
rs770867998 Chr.17: 4167901-4167901 on Build GRCh38
rs574979502 Chr.17: 4167721-4167721 on Build GRCh38
rs757606843 Chr.17: 4167757-4167757 on Build GRCh38
rs932377963 Chr.17: 4167726-4167726 on Build GRCh38
rs189564576 Chr.17: 4167788-4167788 on Build GRCh38
rs750951673 Chr.17: 4167844-4167844 on Build GRCh38
rs149788482 Chr.17: 4167769-4167769 on Build GRCh38
rs200588304 Chr.17: 4167855-4167855 on Build GRCh38
rs1032562042 Chr.17: 4167770-4167770 on Build GRCh38
rs895338421 Chr.17: 4167828-4167828 on Build GRCh38
rs752412370 Chr.17: 4167744-4167744 on Build GRCh38
rs368100646 Chr.17: 4167753-4167753 on Build GRCh38
rs759493361 Chr.17: 4167800-4167800 on Build GRCh38
rs570880275 Chr.17: 4167768-4167768 on Build GRCh38
rs749141442 Chr.17: 4167787-4167787 on Build GRCh38
rs753508116 Chr.17: 4167835-4167835 on Build GRCh38
rs374502871 Chr.17: 4167856-4167856 on Build GRCh38
rs920118010 Chr.17: 4167699-4167699 on Build GRCh38
rs763606938 Chr.17: 4167897-4167897 on Build GRCh38
rs1004930080 Chr.17: 4167863-4167863 on Build GRCh38
rs746343301 Chr.17: 4167767-4167767 on Build GRCh38
rs764528365 Chr.17: 4167752-4167752 on Build GRCh38
rs758836424 Chr.17: 4167848-4167848 on Build GRCh38
rs75366887 Chr.17: 4167681-4167681 on Build GRCh38
rs34065975 Chr.17: 4167765-4167765 on Build GRCh38
rs774799680 Chr.17: 4167829-4167829 on Build GRCh38
rs778789079 Chr.17: 4167902-4167902 on Build GRCh38
rs757231209 Chr.17: 4167876-4167876 on Build GRCh38
rs201367881 Chr.17: 4167762-4167762 on Build GRCh38
rs556607599 Chr.17: 4167779-4167779 on Build GRCh38
rs759154812 Chr.17: 4167728-4167728 on Build GRCh38
rs774457064 Chr.17: 4167790-4167790 on Build GRCh38
rs778206530 Chr.17: 4167768-4167769 on Build GRCh38
rs765547042 Chr.17: 4167842-4167842 on Build GRCh38
rs368284595 Chr.17: 4167786-4167786 on Build GRCh38
rs762207476 Chr.17: 4167839-4167839 on Build GRCh38
rs768825320 Chr.17: 4167774-4167774 on Build GRCh38
rs1009785417 Chr.17: 4167682-4167682 on Build GRCh38
rs921802319 Chr.17: 4167738-4167738 on Build GRCh38
rs377539819 Chr.17: 4167746-4167746 on Build GRCh38
rs568668091 Chr.17: 4167708-4167708 on Build GRCh38
rs371551597 Chr.17: 4167827-4167827 on Build GRCh38
rs375333912 Chr.17: 4167685-4167685 on Build GRCh38
rs767092292 Chr.17: 4167736-4167736 on Build GRCh38
rs781306911 Chr.17: 4167864-4167864 on Build GRCh38
rs763714568 Chr.17: 4167830-4167830 on Build GRCh38

View Full Product Details