Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112267979-112268250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1AR
Entrez Gene ID
55435
Gene Name
adaptor related protein complex 1 associated regulatory protein
Gene Aliases
2C18, C4orf16, GBAR, PRO0971, gamma-BAR
UniGene
Hs.435991
Gene Chromosome Location
Chr.4: 112231727-112273110 on Build GRCh38
Transcripts
NM_001128426.2 NM_018569.5 NR_136622.1 XM_017008378.1 XM_017008379.1 XM_017008380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894479 c.653G>C p.R218T Chr.4: 112268153-112268153 on Build GRCh38
1050095 c.673C>T p.R225C Chr.4: 112268173-112268173 on Build GRCh38
4121386 c.700C>T p.R234W Chr.4: 112268200-112268200 on Build GRCh38
6228426 c.748T>C p.S250P Chr.4: 112268248-112268248 on Build GRCh38
731961 c.672A>G p.E224E Chr.4: 112268172-112268172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000200241 Chr.4: 112268033-112268033 on Build GRCh38
rs375877761 Chr.4: 112268154-112268154 on Build GRCh38
rs761655982 Chr.4: 112268174-112268174 on Build GRCh38
rs775311861 Chr.4: 112268131-112268131 on Build GRCh38
rs913569226 Chr.4: 112268014-112268014 on Build GRCh38
rs776329819 Chr.4: 112268164-112268164 on Build GRCh38
rs745382788 Chr.4: 112268112-112268112 on Build GRCh38
rs1034197181 Chr.4: 112268035-112268035 on Build GRCh38
rs945049723 Chr.4: 112268095-112268095 on Build GRCh38
rs760195199 Chr.4: 112268134-112268134 on Build GRCh38
rs368240881 Chr.4: 112268201-112268201 on Build GRCh38
rs750296615 Chr.4: 112268194-112268194 on Build GRCh38
rs556414510 Chr.4: 112268015-112268015 on Build GRCh38
rs765174258 Chr.4: 112268186-112268186 on Build GRCh38
rs746833115 Chr.4: 112268184-112268185 on Build GRCh38
rs1040972332 Chr.4: 112268200-112268200 on Build GRCh38
rs968815904 Chr.4: 112268012-112268012 on Build GRCh38
rs139102515 Chr.4: 112268091-112268091 on Build GRCh38
rs771638682 Chr.4: 112268129-112268129 on Build GRCh38
rs763053410 Chr.4: 112268198-112268198 on Build GRCh38
rs545718393 Chr.4: 112268203-112268203 on Build GRCh38
rs992655683 Chr.4: 112268082-112268082 on Build GRCh38
rs958758287 Chr.4: 112268058-112268058 on Build GRCh38
rs778406937 Chr.4: 112268111-112268111 on Build GRCh38
rs768332122 Chr.4: 112268155-112268155 on Build GRCh38
rs770268062 Chr.4: 112268094-112268094 on Build GRCh38

View Full Product Details