Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 59312608-59312881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX4
Entrez Gene ID
339302
Gene Name
complexin 4
Gene Aliases
CPX-IV, CPXIV
UniGene
Hs.449884
Gene Chromosome Location
Chr.18: 59295402-59318649 on Build GRCh38
Transcripts
NM_181654.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934153 c.194A>G p.Q65R Chr.18: 59312746-59312746 on Build GRCh38
4674356 c.200delA p.K67fs*44 Chr.18: 59312740-59312740 on Build GRCh38
4072796 c.183T>C p.A61A Chr.18: 59312757-59312757 on Build GRCh38
1202150 c.221G>T p.R74I Chr.18: 59312719-59312719 on Build GRCh38
5388309 c.255G>A p.K85K Chr.18: 59312685-59312685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778682188 Chr.18: 59312704-59312704 on Build GRCh38
rs1044222341 Chr.18: 59312773-59312773 on Build GRCh38
rs752341091 Chr.18: 59312708-59312708 on Build GRCh38
rs371716650 Chr.18: 59312794-59312794 on Build GRCh38
rs754266109 Chr.18: 59312672-59312672 on Build GRCh38
rs914643074 Chr.18: 59312676-59312676 on Build GRCh38
rs752357725 Chr.18: 59312808-59312808 on Build GRCh38
rs780970323 Chr.18: 59312728-59312728 on Build GRCh38
rs777295071 Chr.18: 59312822-59312822 on Build GRCh38
rs892839806 Chr.18: 59312650-59312650 on Build GRCh38
rs760010133 Chr.18: 59312797-59312797 on Build GRCh38
rs749771598 Chr.18: 59312814-59312814 on Build GRCh38
rs755273332 Chr.18: 59312678-59312678 on Build GRCh38
rs113549740 Chr.18: 59312831-59312831 on Build GRCh38
rs765518238 Chr.18: 59312801-59312801 on Build GRCh38
rs766954020 Chr.18: 59312788-59312788 on Build GRCh38
rs774722502 Chr.18: 59312758-59312758 on Build GRCh38
rs146447477 Chr.18: 59312859-59312859 on Build GRCh38
rs199848677 Chr.18: 59312803-59312803 on Build GRCh38
rs762071719 Chr.18: 59312760-59312760 on Build GRCh38
rs777206954 Chr.18: 59312711-59312711 on Build GRCh38
rs372890559 Chr.18: 59312658-59312658 on Build GRCh38
rs772304978 Chr.18: 59312762-59312762 on Build GRCh38
rs770941363 Chr.18: 59312727-59312727 on Build GRCh38
rs200257615 Chr.18: 59312785-59312785 on Build GRCh38
rs375488638 Chr.18: 59312717-59312717 on Build GRCh38
rs112941151 Chr.18: 59312756-59312756 on Build GRCh38
rs766368211 Chr.18: 59312817-59312817 on Build GRCh38
rs1002531017 Chr.18: 59312830-59312830 on Build GRCh38
rs745735567 Chr.18: 59312729-59312729 on Build GRCh38
rs202223771 Chr.18: 59312749-59312749 on Build GRCh38
rs773389769 Chr.18: 59312768-59312768 on Build GRCh38

View Full Product Details