Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67297212-67297483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAND1
Entrez Gene ID
55832
Gene Name
cullin associated and neddylation dissociated 1
Gene Aliases
TIP120, TIP120A
UniGene
Hs.546407
Gene Chromosome Location
Chr.12: 67269281-67314692 on Build GRCh38
Transcripts
NM_018448.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6891113 c.545C>T p.P182L Chr.12: 67297460-67297460 on Build GRCh38
6272931 c.557G>T p.S186I Chr.12: 67297472-67297472 on Build GRCh38
4924886 c.539T>C p.L180P Chr.12: 67297454-67297454 on Build GRCh38
3987168 c.520C>A p.P174T Chr.12: 67297435-67297435 on Build GRCh38
6898277 c.508G>A p.V170I Chr.12: 67297423-67297423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747986387 Chr.12: 67297278-67297278 on Build GRCh38
rs143204578 Chr.12: 67297357-67297357 on Build GRCh38
rs752590541 Chr.12: 67297449-67297449 on Build GRCh38
rs749418936 Chr.12: 67297318-67297318 on Build GRCh38
rs773511417 Chr.12: 67297295-67297295 on Build GRCh38
rs757672317 Chr.12: 67297390-67297394 on Build GRCh38
rs755924756 Chr.12: 67297452-67297452 on Build GRCh38
rs754310023 Chr.12: 67297386-67297387 on Build GRCh38
rs774363546 Chr.12: 67297337-67297337 on Build GRCh38
rs769543467 Chr.12: 67297286-67297286 on Build GRCh38
rs561047488 Chr.12: 67297344-67297344 on Build GRCh38
rs781147405 Chr.12: 67297434-67297434 on Build GRCh38
rs749107015 Chr.12: 67297460-67297460 on Build GRCh38
rs971533402 Chr.12: 67297397-67297397 on Build GRCh38
rs979814155 Chr.12: 67297461-67297461 on Build GRCh38
rs777371200 Chr.12: 67297455-67297455 on Build GRCh38
rs1033181325 Chr.12: 67297354-67297354 on Build GRCh38
rs186043736 Chr.12: 67297266-67297266 on Build GRCh38
rs766047763 Chr.12: 67297393-67297393 on Build GRCh38
rs772572761 Chr.12: 67297240-67297240 on Build GRCh38
rs373853939 Chr.12: 67297364-67297364 on Build GRCh38
rs900792043 Chr.12: 67297281-67297281 on Build GRCh38
rs1047810186 Chr.12: 67297249-67297249 on Build GRCh38
rs566462554 Chr.12: 67297251-67297251 on Build GRCh38
rs751084417 Chr.12: 67297394-67297394 on Build GRCh38
rs764655146 Chr.12: 67297372-67297374 on Build GRCh38
rs754457959 Chr.12: 67297399-67297399 on Build GRCh38
rs764318511 Chr.12: 67297382-67297382 on Build GRCh38
rs750107464 Chr.12: 67297386-67297386 on Build GRCh38
rs761153807 Chr.12: 67297369-67297369 on Build GRCh38
rs376295658 Chr.12: 67297390-67297390 on Build GRCh38

View Full Product Details