Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 98821703-98821976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
UniGene
Hs.469398
Gene Chromosome Location
Chr.2: 98793846-98936221 on Build GRCh38
Transcripts
NM_207362.2 XM_005263923.3 XM_005263924.3 XM_005263926.3 XM_011511093.1 XM_011511094.1 XM_011511095.2 XM_011511096.1 XM_017003971.1 XM_017003972.1 XM_017003973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408173 c.2320C>G p.P774A Chr.2: 98821953-98821953 on Build GRCh38
6048599 c.2339_2340insC p.D781fs*26 Chr.2: 98821933-98821934 on Build GRCh38
5578177 c.2351C>T p.P784L Chr.2: 98821922-98821922 on Build GRCh38
5863066 c.2411G>A p.G804E Chr.2: 98821862-98821862 on Build GRCh38
2153131 c.2383A>G p.T795A Chr.2: 98821890-98821890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527460905 Chr.2: 98821869-98821869 on Build GRCh38
rs772311196 Chr.2: 98821952-98821952 on Build GRCh38
rs368272129 Chr.2: 98821942-98821942 on Build GRCh38
rs999592709 Chr.2: 98821793-98821793 on Build GRCh38
rs765154286 Chr.2: 98821921-98821921 on Build GRCh38
rs924224163 Chr.2: 98821744-98821744 on Build GRCh38
rs374304519 Chr.2: 98821937-98821937 on Build GRCh38
rs752261294 Chr.2: 98821923-98821923 on Build GRCh38
rs548982163 Chr.2: 98821938-98821938 on Build GRCh38
rs749820125 Chr.2: 98821877-98821877 on Build GRCh38
rs549707886 Chr.2: 98821934-98821934 on Build GRCh38
rs753845926 Chr.2: 98821847-98821847 on Build GRCh38
rs760770693 Chr.2: 98821892-98821892 on Build GRCh38
rs375947973 Chr.2: 98821896-98821896 on Build GRCh38
rs1005462198 Chr.2: 98821905-98821905 on Build GRCh38
rs762134940 Chr.2: 98821807-98821807 on Build GRCh38
rs748279818 Chr.2: 98821948-98821948 on Build GRCh38
rs367789387 Chr.2: 98821826-98821826 on Build GRCh38
rs541161975 Chr.2: 98821870-98821870 on Build GRCh38
rs971222863 Chr.2: 98821758-98821758 on Build GRCh38
rs769268167 Chr.2: 98821935-98821935 on Build GRCh38
rs895539404 Chr.2: 98821803-98821803 on Build GRCh38
rs758111492 Chr.2: 98821868-98821868 on Build GRCh38
rs752135060 Chr.2: 98821867-98821867 on Build GRCh38
rs1052788245 Chr.2: 98821783-98821783 on Build GRCh38
rs1044219742 Chr.2: 98821828-98821828 on Build GRCh38
rs375049911 Chr.2: 98821874-98821874 on Build GRCh38
rs767851381 Chr.2: 98821816-98821816 on Build GRCh38
rs911134051 Chr.2: 98821844-98821844 on Build GRCh38
rs774643068 Chr.2: 98821885-98821885 on Build GRCh38
rs370809977 Chr.2: 98821823-98821823 on Build GRCh38
rs778498299 Chr.2: 98821862-98821862 on Build GRCh38
rs1031264913 Chr.2: 98821802-98821802 on Build GRCh38
rs757880548 Chr.2: 98821926-98821926 on Build GRCh38
rs567746539 Chr.2: 98821821-98821821 on Build GRCh38
rs934274857 Chr.2: 98821775-98821775 on Build GRCh38
rs561839252 Chr.2: 98821774-98821774 on Build GRCh38
rs60496162 Chr.2: 98821882-98821882 on Build GRCh38
rs766219898 Chr.2: 98821913-98821913 on Build GRCh38
rs761973568 Chr.2: 98821887-98821887 on Build GRCh38
rs773538963 Chr.2: 98821889-98821889 on Build GRCh38
rs772475114 Chr.2: 98821888-98821888 on Build GRCh38
rs754824478 Chr.2: 98821860-98821860 on Build GRCh38
rs994098527 Chr.2: 98821936-98821936 on Build GRCh38
rs751240763 Chr.2: 98821932-98821932 on Build GRCh38
rs150915151 Chr.2: 98821958-98821958 on Build GRCh38
rs1025092171 Chr.2: 98821755-98821755 on Build GRCh38
rs770162187 Chr.2: 98821873-98821873 on Build GRCh38
rs908065688 Chr.2: 98821895-98821895 on Build GRCh38
rs756806957 Chr.2: 98821933-98821933 on Build GRCh38
rs532793433 Chr.2: 98821915-98821915 on Build GRCh38

View Full Product Details