Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 49279507-49279754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLN1
Entrez Gene ID
869
Gene Name
cerebellin 1 precursor
Gene Aliases
-
UniGene
Hs.458423
Gene Chromosome Location
Chr.16: 49277917-49281831 on Build GRCh38
Transcripts
NM_004352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4060973 c.438G>T p.Q146H Chr.16: 49279548-49279548 on Build GRCh38
6079171 c.398T>G p.L133R Chr.16: 49279588-49279588 on Build GRCh38
5554637 c.432T>C p.G144G Chr.16: 49279554-49279554 on Build GRCh38
3729648 c.457G>A p.A153T Chr.16: 49279529-49279529 on Build GRCh38
5913740 c.431G>A p.G144D Chr.16: 49279555-49279555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773086998 Chr.16: 49279636-49279636 on Build GRCh38
rs534323496 Chr.16: 49279649-49279649 on Build GRCh38
rs992143251 Chr.16: 49279536-49279536 on Build GRCh38
rs543323470 Chr.16: 49279678-49279678 on Build GRCh38
rs768101476 Chr.16: 49279618-49279618 on Build GRCh38
rs373881041 Chr.16: 49279583-49279583 on Build GRCh38
rs747764462 Chr.16: 49279626-49279626 on Build GRCh38
rs771859462 Chr.16: 49279629-49279629 on Build GRCh38
rs377069094 Chr.16: 49279615-49279615 on Build GRCh38
rs752908497 Chr.16: 49279577-49279577 on Build GRCh38
rs557148064 Chr.16: 49279686-49279686 on Build GRCh38
rs747377102 Chr.16: 49279584-49279584 on Build GRCh38
rs929436186 Chr.16: 49279587-49279587 on Build GRCh38
rs1038303738 Chr.16: 49279650-49279650 on Build GRCh38
rs932329795 Chr.16: 49279728-49279728 on Build GRCh38
rs908415607 Chr.16: 49279592-49279592 on Build GRCh38
rs781615579 Chr.16: 49279601-49279601 on Build GRCh38
rs890771599 Chr.16: 49279730-49279730 on Build GRCh38
rs552430485 Chr.16: 49279727-49279727 on Build GRCh38
rs557291520 Chr.16: 49279662-49279662 on Build GRCh38
rs755940159 Chr.16: 49279560-49279561 on Build GRCh38
rs760601519 Chr.16: 49279648-49279648 on Build GRCh38
rs773936747 Chr.16: 49279620-49279620 on Build GRCh38
rs765465461 Chr.16: 49279575-49279575 on Build GRCh38
rs111507068 Chr.16: 49279726-49279727 on Build GRCh38
rs758640384 Chr.16: 49279580-49279580 on Build GRCh38
rs1050809601 Chr.16: 49279729-49279729 on Build GRCh38
rs757749509 Chr.16: 49279591-49279591 on Build GRCh38
rs1009351318 Chr.16: 49279733-49279733 on Build GRCh38
rs577885911 Chr.16: 49279669-49279669 on Build GRCh38
rs777637020 Chr.16: 49279561-49279562 on Build GRCh38
rs975452093 Chr.16: 49279691-49279691 on Build GRCh38

View Full Product Details