Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7041787-7042042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC96
Entrez Gene ID
257236
Gene Name
coiled-coil domain containing 96
Gene Aliases
-
UniGene
Hs.656757
Gene Chromosome Location
Chr.4: 7040849-7043001 on Build GRCh38
Transcripts
NM_153376.2 NR_033828.1

Gene Symbol
LOC100129931
Entrez Gene ID
100129931
Gene Name
uncharacterized LOC100129931
Gene Aliases
-
UniGene
Hs.707753
Gene Chromosome Location
Chr.4: 7030554-7046231 on Build GRCh38
Transcripts
NM_153376.2 NR_033828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622721 c.1095G>A p.E365E Chr.4: 7041844-7041844 on Build GRCh38
2829744 c.1127G>T p.R376L Chr.4: 7041812-7041812 on Build GRCh38
6228028 c.1021G>A p.A341T Chr.4: 7041918-7041918 on Build GRCh38
5060437 c.1089G>A p.A363A Chr.4: 7041850-7041850 on Build GRCh38
7324393 c.1029C>T p.G343G Chr.4: 7041910-7041910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200146451 Chr.4: 7041999-7041999 on Build GRCh38
rs746179622 Chr.4: 7041891-7041891 on Build GRCh38
rs750814948 Chr.4: 7041879-7041880 on Build GRCh38
rs772714839 Chr.4: 7041913-7041913 on Build GRCh38
rs759446021 Chr.4: 7041964-7041964 on Build GRCh38
rs781421287 Chr.4: 7041890-7041890 on Build GRCh38
rs771552044 Chr.4: 7041912-7041912 on Build GRCh38
rs143021792 Chr.4: 7041819-7041819 on Build GRCh38
rs762943312 Chr.4: 7042000-7042000 on Build GRCh38
rs757439679 Chr.4: 7042024-7042024 on Build GRCh38
rs201982722 Chr.4: 7041871-7041871 on Build GRCh38
rs770570997 Chr.4: 7041933-7041933 on Build GRCh38
rs752650608 Chr.4: 7041873-7041873 on Build GRCh38
rs372041606 Chr.4: 7041956-7041956 on Build GRCh38
rs148175400 Chr.4: 7041867-7041867 on Build GRCh38
rs200832707 Chr.4: 7041812-7041812 on Build GRCh38
rs917833815 Chr.4: 7041872-7041872 on Build GRCh38
rs773885918 Chr.4: 7041816-7041816 on Build GRCh38
rs758600026 Chr.4: 7041879-7041879 on Build GRCh38
rs377449312 Chr.4: 7041851-7041851 on Build GRCh38
rs760374259 Chr.4: 7041826-7041826 on Build GRCh38
rs764084810 Chr.4: 7042005-7042005 on Build GRCh38
rs775061490 Chr.4: 7041828-7041828 on Build GRCh38
rs765102556 Chr.4: 7041972-7041972 on Build GRCh38
rs200608575 Chr.4: 7041848-7041848 on Build GRCh38
rs1044735081 Chr.4: 7041882-7041882 on Build GRCh38
rs776600923 Chr.4: 7041953-7041953 on Build GRCh38
rs780575039 Chr.4: 7041897-7041897 on Build GRCh38
rs757431893 Chr.4: 7041889-7041889 on Build GRCh38
rs574868062 Chr.4: 7041880-7041880 on Build GRCh38
rs964232832 Chr.4: 7041965-7041965 on Build GRCh38
rs369900664 Chr.4: 7041903-7041903 on Build GRCh38
rs751597363 Chr.4: 7042019-7042019 on Build GRCh38
rs760332684 Chr.4: 7041829-7041831 on Build GRCh38
rs984248342 Chr.4: 7041830-7041830 on Build GRCh38
rs746656628 Chr.4: 7041915-7041915 on Build GRCh38
rs751794110 Chr.4: 7041881-7041881 on Build GRCh38
rs141992507 Chr.4: 7041973-7041973 on Build GRCh38
rs775626492 Chr.4: 7041989-7041989 on Build GRCh38
rs771575015 Chr.4: 7041817-7041817 on Build GRCh38
rs1043563888 Chr.4: 7041866-7041866 on Build GRCh38
rs770222506 Chr.4: 7041814-7041814 on Build GRCh38
rs564940150 Chr.4: 7041832-7041834 on Build GRCh38

View Full Product Details