Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109974256-109974523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV1
Entrez Gene ID
27012
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
HNKA, KCNB3, KV2.3, KV8.1
UniGene
Hs.13285
Gene Chromosome Location
Chr.8: 109963645-109975847 on Build GRCh38
Transcripts
NM_014379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533146 c.94G>A p.G32R Chr.8: 109974295-109974295 on Build GRCh38
6355831 c.84C>T p.S28S Chr.8: 109974305-109974305 on Build GRCh38
6428217 c.16A>T p.R6* Chr.8: 109974373-109974373 on Build GRCh38
4651618 c.111C>T p.L37L Chr.8: 109974278-109974278 on Build GRCh38
7187225 c.132C>A p.N44K Chr.8: 109974257-109974257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961500345 Chr.8: 109974290-109974290 on Build GRCh38
rs948391012 Chr.8: 109974332-109974332 on Build GRCh38
rs544471606 Chr.8: 109974492-109974492 on Build GRCh38
rs530459400 Chr.8: 109974471-109974471 on Build GRCh38
rs34461230 Chr.8: 109974341-109974342 on Build GRCh38
rs900574212 Chr.8: 109974497-109974497 on Build GRCh38
rs7822437 Chr.8: 109974503-109974503 on Build GRCh38
rs1053697301 Chr.8: 109974418-109974418 on Build GRCh38
rs1046900437 Chr.8: 109974280-109974280 on Build GRCh38
rs745331383 Chr.8: 109974341-109974341 on Build GRCh38
rs954455651 Chr.8: 109974494-109974494 on Build GRCh38
rs775731785 Chr.8: 109974300-109974300 on Build GRCh38
rs769233820 Chr.8: 109974380-109974380 on Build GRCh38
rs529769410 Chr.8: 109974415-109974415 on Build GRCh38
rs929951079 Chr.8: 109974449-109974449 on Build GRCh38
rs564518657 Chr.8: 109974464-109974464 on Build GRCh38
rs900328853 Chr.8: 109974485-109974485 on Build GRCh38
rs906648399 Chr.8: 109974334-109974334 on Build GRCh38
rs936231087 Chr.8: 109974391-109974391 on Build GRCh38
rs572839882 Chr.8: 109974325-109974325 on Build GRCh38
rs984370595 Chr.8: 109974277-109974277 on Build GRCh38
rs1047067855 Chr.8: 109974457-109974457 on Build GRCh38
rs926210428 Chr.8: 109974389-109974389 on Build GRCh38
rs916729786 Chr.8: 109974275-109974275 on Build GRCh38
rs17852611 Chr.8: 109974324-109974324 on Build GRCh38
rs1006102702 Chr.8: 109974465-109974465 on Build GRCh38
rs892001194 Chr.8: 109974447-109974447 on Build GRCh38
rs181155062 Chr.8: 109974498-109974498 on Build GRCh38
rs377263756 Chr.8: 109974278-109974278 on Build GRCh38
rs916990543 Chr.8: 109974309-109974309 on Build GRCh38
rs996045505 Chr.8: 109974487-109974487 on Build GRCh38
rs992612607 Chr.8: 109974308-109974308 on Build GRCh38
rs779307333 Chr.8: 109974274-109974274 on Build GRCh38
rs545133669 Chr.8: 109974377-109974377 on Build GRCh38

View Full Product Details