Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45535119-45535376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
UniGene
Hs.24601
Gene Chromosome Location
Chr.22: 45502839-45601134 on Build GRCh38
Transcripts
NM_001996.3 NM_006485.3 NM_006486.2 NM_006487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
275131 c.826G>A p.D276N Chr.22: 45535241-45535241 on Build GRCh38
4634814 c.807T>C p.G269G Chr.22: 45535222-45535222 on Build GRCh38
1035078 c.804T>C p.S268S Chr.22: 45535219-45535219 on Build GRCh38
6571629 c.907C>G p.L303V Chr.22: 45535322-45535322 on Build GRCh38
3973014 c.869C>A p.P290H Chr.22: 45535284-45535284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111292144 Chr.22: 45535343-45535343 on Build GRCh38
rs774051379 Chr.22: 45535354-45535354 on Build GRCh38
rs764616641 Chr.22: 45535278-45535278 on Build GRCh38
rs527820999 Chr.22: 45535154-45535154 on Build GRCh38
rs749250097 Chr.22: 45535352-45535352 on Build GRCh38
rs35051354 Chr.22: 45535287-45535288 on Build GRCh38
rs770350150 Chr.22: 45535240-45535240 on Build GRCh38
rs147740669 Chr.22: 45535351-45535351 on Build GRCh38
rs754315869 Chr.22: 45535218-45535218 on Build GRCh38
rs750631703 Chr.22: 45535315-45535315 on Build GRCh38
rs551195879 Chr.22: 45535276-45535276 on Build GRCh38
rs768437295 Chr.22: 45535174-45535174 on Build GRCh38
rs759317468 Chr.22: 45535241-45535241 on Build GRCh38
rs758669185 Chr.22: 45535326-45535326 on Build GRCh38
rs138580627 Chr.22: 45535297-45535297 on Build GRCh38
rs753148981 Chr.22: 45535275-45535275 on Build GRCh38
rs757752021 Chr.22: 45535301-45535301 on Build GRCh38
rs781024644 Chr.22: 45535188-45535188 on Build GRCh38
rs780934807 Chr.22: 45535330-45535330 on Build GRCh38
rs890297116 Chr.22: 45535284-45535284 on Build GRCh38
rs144704783 Chr.22: 45535207-45535207 on Build GRCh38
rs200333117 Chr.22: 45535312-45535312 on Build GRCh38
rs926390338 Chr.22: 45535189-45535189 on Build GRCh38
rs892485260 Chr.22: 45535169-45535169 on Build GRCh38
rs757309364 Chr.22: 45535158-45535158 on Build GRCh38
rs945288675 Chr.22: 45535247-45535247 on Build GRCh38
rs762466706 Chr.22: 45535236-45535236 on Build GRCh38
rs746097365 Chr.22: 45535164-45535164 on Build GRCh38
rs754317939 Chr.22: 45535291-45535291 on Build GRCh38
rs773046273 Chr.22: 45535224-45535224 on Build GRCh38
rs747898197 Chr.22: 45535332-45535332 on Build GRCh38
rs753919551 Chr.22: 45535157-45535157 on Build GRCh38
rs113285528 Chr.22: 45535197-45535197 on Build GRCh38
rs981872203 Chr.22: 45535155-45535155 on Build GRCh38
rs767967831 Chr.22: 45535274-45535274 on Build GRCh38
rs761060703 Chr.22: 45535175-45535175 on Build GRCh38
rs903812631 Chr.22: 45535252-45535252 on Build GRCh38
rs531089378 Chr.22: 45535194-45535194 on Build GRCh38
rs944584555 Chr.22: 45535294-45535294 on Build GRCh38

View Full Product Details