Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66566014-66566283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
UniGene
Hs.11590
Gene Chromosome Location
Chr.11: 66563463-66568576 on Build GRCh38
Transcripts
NM_003793.3 XM_011545328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6552310 c.803G>A p.G268D Chr.11: 66566086-66566086 on Build GRCh38
147276 c.722-5C>T p.? Chr.11: 66566172-66566172 on Build GRCh38
6552311 c.795G>A p.K265K Chr.11: 66566094-66566094 on Build GRCh38
689886 c.727G>A p.E243K Chr.11: 66566162-66566162 on Build GRCh38
7116988 c.727_729delGAG p.E243delE Chr.11: 66566160-66566162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776395150 Chr.11: 66566052-66566052 on Build GRCh38
rs763662917 Chr.11: 66566150-66566150 on Build GRCh38
rs745821135 Chr.11: 66566049-66566050 on Build GRCh38
rs756264326 Chr.11: 66566101-66566101 on Build GRCh38
rs1026587067 Chr.11: 66566229-66566229 on Build GRCh38
rs374695833 Chr.11: 66566253-66566253 on Build GRCh38
rs563045234 Chr.11: 66566156-66566156 on Build GRCh38
rs910288326 Chr.11: 66566098-66566098 on Build GRCh38
rs769735423 Chr.11: 66566160-66566162 on Build GRCh38
rs780325681 Chr.11: 66566171-66566171 on Build GRCh38
rs963888266 Chr.11: 66566205-66566205 on Build GRCh38
rs140002533 Chr.11: 66566109-66566109 on Build GRCh38
rs762614811 Chr.11: 66566146-66566146 on Build GRCh38
rs769512466 Chr.11: 66566128-66566128 on Build GRCh38
rs186275453 Chr.11: 66566243-66566243 on Build GRCh38
rs748923631 Chr.11: 66566246-66566246 on Build GRCh38
rs764869338 Chr.11: 66566056-66566056 on Build GRCh38
rs775268694 Chr.11: 66566137-66566137 on Build GRCh38
rs980671668 Chr.11: 66566046-66566046 on Build GRCh38
rs779758651 Chr.11: 66566195-66566197 on Build GRCh38
rs767931300 Chr.11: 66566155-66566155 on Build GRCh38
rs894543359 Chr.11: 66566182-66566182 on Build GRCh38
rs201753663 Chr.11: 66566153-66566153 on Build GRCh38
rs147398226 Chr.11: 66566079-66566079 on Build GRCh38
rs573169893 Chr.11: 66566204-66566205 on Build GRCh38
rs746899739 Chr.11: 66566111-66566111 on Build GRCh38
rs773124981 Chr.11: 66566037-66566037 on Build GRCh38
rs941653140 Chr.11: 66566076-66566076 on Build GRCh38
rs371200527 Chr.11: 66566108-66566108 on Build GRCh38
rs755101005 Chr.11: 66566180-66566180 on Build GRCh38
rs767970620 Chr.11: 66566262-66566262 on Build GRCh38
rs954969337 Chr.11: 66566237-66566237 on Build GRCh38
rs138591048 Chr.11: 66566209-66566209 on Build GRCh38
rs752186392 Chr.11: 66566058-66566058 on Build GRCh38
rs561399021 Chr.11: 66566236-66566236 on Build GRCh38
rs762399053 Chr.11: 66566260-66566260 on Build GRCh38
rs781386282 Chr.11: 66566086-66566086 on Build GRCh38
rs752001068 Chr.11: 66566078-66566078 on Build GRCh38
rs763567392 Chr.11: 66566059-66566059 on Build GRCh38
rs932976138 Chr.11: 66566065-66566065 on Build GRCh38
rs759062690 Chr.11: 66566055-66566055 on Build GRCh38
rs747001135 Chr.11: 66566208-66566208 on Build GRCh38
rs879701204 Chr.11: 66566181-66566181 on Build GRCh38
rs545009 Chr.11: 66566127-66566127 on Build GRCh38
rs78871927 Chr.11: 66566172-66566172 on Build GRCh38
rs781052937 Chr.11: 66566242-66566242 on Build GRCh38
rs764755120 Chr.11: 66566073-66566073 on Build GRCh38
rs368626946 Chr.11: 66566097-66566097 on Build GRCh38
rs780124294 Chr.11: 66566102-66566102 on Build GRCh38
rs375983534 Chr.11: 66566191-66566191 on Build GRCh38
rs879899565 Chr.11: 66566139-66566139 on Build GRCh38
rs182076457 Chr.11: 66566211-66566211 on Build GRCh38
rs760584939 Chr.11: 66566040-66566040 on Build GRCh38
rs970950102 Chr.11: 66566230-66566230 on Build GRCh38
rs773768971 Chr.11: 66566264-66566264 on Build GRCh38
rs769707236 Chr.11: 66566244-66566244 on Build GRCh38
rs776247858 Chr.11: 66566119-66566119 on Build GRCh38
rs773692685 Chr.11: 66566255-66566255 on Build GRCh38
rs1016425852 Chr.11: 66566203-66566203 on Build GRCh38
rs756460539 Chr.11: 66566158-66566158 on Build GRCh38

View Full Product Details