Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73959117-73959372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ6
Entrez Gene ID
51004
Gene Name
coenzyme Q6, monooxygenase
Gene Aliases
CGI-10, CGI10, COQ10D6
UniGene
Hs.131555
Gene Chromosome Location
Chr.14: 73949934-73963112 on Build GRCh38
Transcripts
NM_182476.2 NM_182480.2 XM_006720156.1 XM_011536807.1 XM_011536808.1 XM_011536809.2 XM_011536810.2 XM_017021351.1 XM_017021352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6030664 c.764G>T p.G255V Chr.14: 73959205-73959205 on Build GRCh38
6634154 c.738A>G p.V246V Chr.14: 73959179-73959179 on Build GRCh38
6288315 c.778C>T p.L260F Chr.14: 73959219-73959219 on Build GRCh38
957707 c.771T>C p.I257I Chr.14: 73959212-73959212 on Build GRCh38
957706 c.730_732delAAC p.N245delN Chr.14: 73959171-73959173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371260604 Chr.14: 73959223-73959223 on Build GRCh38
rs576411727 Chr.14: 73959274-73959274 on Build GRCh38
rs1029775676 Chr.14: 73959257-73959257 on Build GRCh38
rs761536538 Chr.14: 73959184-73959184 on Build GRCh38
rs775053324 Chr.14: 73959161-73959161 on Build GRCh38
rs773923049 Chr.14: 73959330-73959330 on Build GRCh38
rs772524163 Chr.14: 73959259-73959259 on Build GRCh38
rs761630783 Chr.14: 73959334-73959334 on Build GRCh38
rs959667329 Chr.14: 73959234-73959234 on Build GRCh38
rs140822482 Chr.14: 73959219-73959219 on Build GRCh38
rs780163765 Chr.14: 73959221-73959221 on Build GRCh38
rs751338512 Chr.14: 73959177-73959177 on Build GRCh38
rs759004340 Chr.14: 73959311-73959311 on Build GRCh38
rs915624002 Chr.14: 73959249-73959249 on Build GRCh38
rs539213991 Chr.14: 73959216-73959216 on Build GRCh38
rs373668656 Chr.14: 73959151-73959151 on Build GRCh38
rs200918369 Chr.14: 73959176-73959176 on Build GRCh38
rs748537586 Chr.14: 73959248-73959248 on Build GRCh38
rs758998842 Chr.14: 73959149-73959149 on Build GRCh38
rs951290557 Chr.14: 73959210-73959210 on Build GRCh38
rs747528224 Chr.14: 73959290-73959290 on Build GRCh38
rs1017183104 Chr.14: 73959203-73959203 on Build GRCh38
rs769227613 Chr.14: 73959295-73959295 on Build GRCh38
rs762634372 Chr.14: 73959175-73959175 on Build GRCh38
rs141046392 Chr.14: 73959332-73959332 on Build GRCh38
rs146605584 Chr.14: 73959145-73959145 on Build GRCh38
rs779480762 Chr.14: 73959225-73959225 on Build GRCh38
rs769439302 Chr.14: 73959282-73959283 on Build GRCh38
rs199768730 Chr.14: 73959224-73959224 on Build GRCh38
rs767443437 Chr.14: 73959195-73959195 on Build GRCh38
rs1003871804 Chr.14: 73959222-73959222 on Build GRCh38
rs766437478 Chr.14: 73959345-73959345 on Build GRCh38
rs773077068 Chr.14: 73959336-73959336 on Build GRCh38
rs969608411 Chr.14: 73959255-73959255 on Build GRCh38
rs750338458 Chr.14: 73959213-73959213 on Build GRCh38
rs775056983 Chr.14: 73959327-73959327 on Build GRCh38
rs760633667 Chr.14: 73959343-73959343 on Build GRCh38
rs747777146 Chr.14: 73959267-73959268 on Build GRCh38
rs899683041 Chr.14: 73959171-73959171 on Build GRCh38
rs1057519350 Chr.14: 73959204-73959204 on Build GRCh38
rs780694613 Chr.14: 73959238-73959242 on Build GRCh38
rs773728803 Chr.14: 73959309-73959309 on Build GRCh38

View Full Product Details