Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24301531-24301800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
UniGene
Hs.708040
Gene Chromosome Location
Chr.14: 24299854-24309126 on Build GRCh38
Transcripts
NM_001286367.1 NM_174913.2 XM_005267385.1 XM_011536526.1 XM_011536527.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420282 c.794T>C p.L265P Chr.14: 24301708-24301708 on Build GRCh38
6056106 c.770T>G p.L257R Chr.14: 24301684-24301684 on Build GRCh38
1660837 c.706A>T p.K236* Chr.14: 24301620-24301620 on Build GRCh38
6298135 c.700G>T p.A234S Chr.14: 24301614-24301614 on Build GRCh38
4751368 c.769C>A p.L257I Chr.14: 24301683-24301683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368599863 Chr.14: 24301578-24301578 on Build GRCh38
rs781233063 Chr.14: 24301610-24301610 on Build GRCh38
rs761078837 Chr.14: 24301580-24301580 on Build GRCh38
rs748958322 Chr.14: 24301671-24301671 on Build GRCh38
rs750470503 Chr.14: 24301599-24301599 on Build GRCh38
rs762754507 Chr.14: 24301751-24301751 on Build GRCh38
rs754906397 Chr.14: 24301602-24301602 on Build GRCh38
rs187305485 Chr.14: 24301585-24301585 on Build GRCh38
rs768416000 Chr.14: 24301697-24301699 on Build GRCh38
rs972097299 Chr.14: 24301668-24301668 on Build GRCh38
rs761166852 Chr.14: 24301716-24301716 on Build GRCh38
rs778876088 Chr.14: 24301591-24301591 on Build GRCh38
rs143004628 Chr.14: 24301680-24301680 on Build GRCh38
rs746491577 Chr.14: 24301565-24301565 on Build GRCh38
rs371242707 Chr.14: 24301729-24301729 on Build GRCh38
rs756036876 Chr.14: 24301618-24301618 on Build GRCh38
rs766559764 Chr.14: 24301750-24301750 on Build GRCh38
rs772929760 Chr.14: 24301566-24301566 on Build GRCh38
rs761967030 Chr.14: 24301724-24301724 on Build GRCh38
rs776927069 Chr.14: 24301723-24301723 on Build GRCh38
rs570256840 Chr.14: 24301705-24301705 on Build GRCh38
rs146695481 Chr.14: 24301669-24301669 on Build GRCh38
rs147768520 Chr.14: 24301712-24301712 on Build GRCh38
rs113260348 Chr.14: 24301761-24301761 on Build GRCh38
rs371665435 Chr.14: 24301757-24301757 on Build GRCh38
rs963924036 Chr.14: 24301619-24301619 on Build GRCh38
rs765463057 Chr.14: 24301731-24301731 on Build GRCh38
rs776147405 Chr.14: 24301730-24301730 on Build GRCh38
rs770568914 Chr.14: 24301673-24301673 on Build GRCh38
rs1008260371 Chr.14: 24301593-24301593 on Build GRCh38
rs970803772 Chr.14: 24301703-24301703 on Build GRCh38
rs758561054 Chr.14: 24301747-24301747 on Build GRCh38
rs960167422 Chr.14: 24301647-24301647 on Build GRCh38
rs748081782 Chr.14: 24301614-24301614 on Build GRCh38
rs937180729 Chr.14: 24301737-24301737 on Build GRCh38
rs768027387 Chr.14: 24301573-24301573 on Build GRCh38
rs371906510 Chr.14: 24301581-24301581 on Build GRCh38
rs367862947 Chr.14: 24301741-24301741 on Build GRCh38
rs777733136 Chr.14: 24301772-24301772 on Build GRCh38
rs374811513 Chr.14: 24301588-24301588 on Build GRCh38
rs745421528 Chr.14: 24301681-24301681 on Build GRCh38
rs928497238 Chr.14: 24301713-24301713 on Build GRCh38
rs764447726 Chr.14: 24301722-24301722 on Build GRCh38

View Full Product Details